|
Air-Sensitive Glassware
Air-Sensitive Septa
|
Schlenk-Type • Storage • Septa • Inert Gases & Equipment
Septa Accessories
On this page: Natural rubber • Serrated rubber • Serrated silicone rubber
Natural rubber septa
|
For use with:
|
White rubber Catalog Number
|
Red rubber Catalog Number
|
|
11. 5mm NMR tubes
|
12. 7mm o.d. glass tubing
|
13. 8 or 9mm o.d. glass tubing
|
14. 9 or 10mm o.d. glass tubing
|
15. 13mm i.d. tubing
|
16. S.T. 10/30 joints and small test tubes
|
|
|
| 17. S.T. 14/20 joints* |
|
|
|
18. S.T. 19/22 joints**
|
|
|
| 19. S.T. 24/40 joints*** |
|
|
| 10. Mixed set, 145 pieces with carryingbox, contains the following: 25 each of 1 to 3 above, 20 each of 4 and 6 above, 10 each of 7 to 9 above |
|
|
* Also fits Aldrich 2, 4, and 8oz narrow-mouth bottles. ** Also fits Aldrich 10, 32, 40oz, and 1L narrow-mouth bottles, and 125mL and 1L Sure/Seal bottles. *** Also fits Aldrich 64, 80oz, and 1gal narrow-mouth bottles.
Back to Top
Serrated rubber septa
 |
Closure has a turnover flange molded to grip outside of container neck, promoting a double seal in conjunction with annular serrations on a hollow plug. Annular serrations depress themselves against the inside wall of the bottle neck, making each serration a suction sealing point.
|
|
Fits container neck i.d. (mm)
|
For use with:
|
White rubber Catalog Number
|
Red rubber Catalog Number
|
|
8.0
|
10/30
|
12.5
|
14/20
|
16.0
|
Sure/Seal
|
17.5
|
19/22
|
20.5
|
24/40
|
25.5
|
29/42
|
|
|
Back to Top
Serrated silicone rubber septa
Translucent silicone rubber septa that contains no pigments, waxes, or molding agents.
|
|
Fits container neck i.d. (mm)
|
For use with:
|
White rubber Catalog Number
|
|
8.0
|
10/30
|
|
|
12.5
|
14/20
|
|
|
16.0
|
Sure/Seal
|
|
|
17.5
|
19/22
|
|
|
20.5
|
24/40
|
|
|
25.5
|
29/42