|
Air-Sensitive Glassware
Schlenk Accessories
|
|
|
Introduction • Typical Operations • Accessories
Aldrich Schlenk-Type Glassware Product Accesories
On this page: Deoxygenation apparatus • Schlaker Reaction Tubes • Firestone valve
Deoxygenation apparatus
 |
One-piece condenser/flask unit for degassing solvents. Flask has angled Standard Taper joint and 7mm gas dispersion tube (porosity 25-50 mm) with 2mm glass stopcock. 250mL flask capacity, S.T. 14/20 joint.
Z22,046-9
|
Back to Top
Schlaker Reaction Tubes
 |
 |
The glass frit in the side-arm permits filtration of excess reagents and solvent removal with complete recovery of the resin for the next reaction. Made of heavy-wall glass, with 2mm glass stopcock and frit (porosity 25-50 mm) on side-arm, and a plain or a threaded S.T 14/20 top joint. Compatible with Schlenk glassware and with the Aldrich All-in-One Glassware kit.
|
Schlaker Tube Technique
- Place resin in the tube.
- Evacuate and purge with N2 to remove oxygen via a rubber septum or a septum-inlet adapter in the top joint.
- Stir gently while heating in an oil bath for high temperatures, or place on a shaker for room temperature reactions.
- When the reaction is complete, apply positive N2 pressure through the septum in the top joint, and vacuum to the side-arm.
- Tilt the Schlaker tube on its side to allow solvent and excess reagents to flow out, leaving the resin behind.
|
Cap. (mL)
|
S.T. 14/20 joint Catalog Number
|
Screw-thread joint Catalog Number
|
|
10
|
25
|
50
|
100
|
250
|
|
|
Back to Top
Firestone valve
 |
A rapid, efficient, and foolproof purge valve for 100% replacement of air in reaction vessels. Automatically controls gas flow and pressure. No special fittings needed. Connect reaction vessel, house-vacuum, and purge gas to the Firestone valve via 10mm o.d. connections. Conserve expensive gases by decreasing flow to almost zero after purging.
Z10,361-6
|
See the Flasks Section for a full listing of standard taper schlenk flasks. |