Glassware Catalog
Bubblers - Sigma-Aldrich Glassware Catalog
On this page:
Mini gas bubblerReaction vessel bubblerIn-line bubblerReverse-flow bubblerIn-line oil bubblersCheck-valve bubblers • Safety bubblerMercury bubblerAccessories
Mini gas bubbler
For bubble counting.
Capacity 4mL.
Z223719
Back to Top
Reaction vessel bubbler
Capacity 5 to 7mL.
S. T. Joint
Catalog Number
14/20
19/22
24/40
29/32
Back to Top
In-line bubbler
Capacity 5mL to 7mL.
Z101214
Back to Top
Reverse-flow bubbler
Monitors gas flow into (or gas evolution from) a system.
Capacity 25mL per chamber.
Z147958
Back to Top
In-line oil bubblers
Connect reaction vessel to in-line S.T. joint or use with a ballast bulb to keep pressure constant. Capacity 10mL.
S.T. Joint
Catalog Number
14/20
19/22
19/38
24/40
29/32
Back to Top
Check-valve bubblers
Check-valve ball prevents oil from being drawn into system.
Capacity 15mL.
Description
Catalog Number
Single-inlet tube, top outlet
T-inlet tube, side outlet
Back to Top
Safety bubbler
Built-in flash arrester bulb prevents backflow.
Capacity 15mL fill mark.
Z223727
Back to Top
Mercury bubbler
Permits increased pressure within reaction vessels. Internal reservoir prevents mercury from being drawn into reaction flask or pushed our of bubbler. Do not overfill with mercury. S.T. 24/40 joint.
Capacity 40mL.
Z152439
Mineral Oil
6 x12mL
161403
Back to Top
Accessories
Tygon (Formula 2075) chemically resistant tubing clear, flexible tubing suitable for specialty chemical production and hazardous material handling. Autoclavable.
  • Resistant to MEK, acids, bases, ketones, and alcohols
  • Plasticizer free no contamination from leaching
  • Biocompatible meets USP Class VI criteria
  • No disposal concerns releases only carbon dioxide and water upon incineration
I.d. (in.)
Wall (in.)
O.d. (in.)
Catalog Number
1/16
1/16
3/16
1/8
1/16
1/4
3/16
1/16
5/16
1/4
1/16
3/8
5/16
1/16
7/16
3/8
1/16
1/2
1/2
1/16
3/4
5/8
1/8
7/8