|
Glassware Catalog
Distillation Column Packing
|
On this page: Aldrich glass helices • PRO-PAK distillation packings Solid-borosilicate glass beads • PTFE boiling stones
Aldrich glass helices
 |
Each helice consists of a single turn of borosilicate glass rod, approximately 0.5mm in diam.
|
Approx. coil i.d. (mm)
|
Catalog Number
|
|
2
|
|
|
3
|
|
|
4-5
|
|
|
6
|
|
|
Back to top
PRO-PAK distillation packings
Efficient, self-wetting, protruded metal dump-packing for distillation columns of 0.75 to 12in. diameter. Maximum reflux rate of approximately 3.4L/h. Free space for 0.16in.2 size is 94% and for 0.24in.2 size is 96%. Each package has a packing volume of approx. 1L.
|
Metal
|
0.16in.2 Catalog Number
|
0.24in.2 Catalog Number
|
| 316 SS |
|
|
| Monel |
|
|
Back to top
Solid-borosilicate glass beads
 |
For use as distillation column packing, mixing beads, and boiling stones. Each 454g box has a packing volume of approx. 360cm3.
|
Diam. (mm)
|
Catalog Number
|
|
1
|
|
|
2
|
|
|
3
|
|
|
4
|
|
|
5
|
|
|
6
|
|
|
Back to top
PTFE boiling stones
 |
|
PTFE stones are not affected by boiling acids or alkalis, ketones, esters, or alcohols. Easily cleaned and will not scratch glassware.
Z24,355-8