|
Glassware Products
Distillation Traps
|
Dean-Stark Traps • Cold-Traps & Vacuum Traps
On this page: Dean-Stark traps • Jacketed Dean-Stark trap • Modified Dean-Stark apparatus Modified, jacketed Dean-Stark trap • "N"-tubes
Dean-Stark traps
 |
Designed specifically for azeotropic distillation using solvents lighter than water. Each Dean-Stark trap comes standard with a 4mm bore PTFE stopcock. The 12mL and 25mL traps use a 2mm bore stopcocks.
 |
|
Cap. (mL)
|
Height (mm)
|
Side-arm clearance (mm)
|
Suggested flask size
|
S.T. 14/20 Joints Catalog Number
|
S.T. 24/40 Joints Catalog Number
|
S.T. 29/32 Joints Catalog Number
|
|
12
|
205
|
80
|
50mL - 5L
|
|
-
|
-
|
|
25
|
255
|
80
|
50mL - 5L
|
|
-
|
-
|
|
25
|
280
|
80
|
50mL - 5L
|
-
|
100
|
350
|
100
|
5L - 12L
|
-
|
300
|
440
|
170
|
22L - 50L
|
-
|
500
|
455
|
170
|
22L - 50L
|
-
|
|
|
Back to top
Jacketed Dean-Stark trap
For azeotroping large amounts of water, 325mL capacity. With S.T. 24/40 joints and 4mm bore PTFE stopcock.
Z10,257-1
Back to top
Modified Dean-Stark apparatus
|
|
The 8mL reservoir version is specifically designed for small scale azeotropic distillation of valuable solvents using heavy water. 2mm bore PTFE stopcock, 300mm overall height.
|
 |
The modified Dean-Stark technique (See Fig. 1)
- Solvent vapors condense and collect in the receiver
- The modified Dean-Stark trap separates the two components
- The heavier than water layer sinks to the bottom of the trap leaving the water on top
- The solvent is forced back through the inside tube and returned to the boiling flask
- The upper water layer is drained out through the 2mm PTFE stopcock
|
|
Reservoir cap. (mL)
|
S.T. Joint 24/40 Catalog Number
|
S.T. Joint 29/32 Catalog Number
|
|
8
|
35
|
|
|
Back to top
Modified, jacketed Dean-Stark trap
 |
|
350mL capacity. 0-3mm PTFE valve. 300mm overall height.
Z24,960-2 |
Back to top
"N"-tubes
 |
For small-scale distillation. 10mL capacity.
|
Description
|
Catalog Number
|
| S.T. 19/38F, 24/40M joints |
|
| S.T. 24/40 joints |