Glassware Catalog
Evaporators &Condensers - Sigma-Aldrich Glassware Catalog
Glassware  •  Condensers  •  Flasks
On this page:
Aldrich rotary evaporator diagonal condenserDry ice condensers
Dry ice condenser-trapBüchi condensers
Aldrich evaporator condensers fit Büchi rotary evaporator models R110, R114, R124 and R151.
Aldrich rotary evaporator diagonal condenser
35/25 side ball joint at 105° angle, and No. 50 flange. 370mm overall length.
Z22,477-4
Back to top
Dry ice condenser
For use with short-stem duct, 35/20 ball joint
Description
Catalog Number
Trap with hose barb
Trap with a 4mm high vacuum glass stopcock
Dry ice condenser
35/20 ball joint.
Z15,244-7
Dry ice condenser
With S.T. 24/40 ground-glass joints. For use with dry-ice/acetone slush or liquid nitrogen in cold-finger. 75mm o.d., 570mm H.
Z10,309-8
Back to top
Dry ice condenser-trap
Use with or without dry ice to condense and collect in trap. 4mm PTFE stopcock, 85mm o.d.
Reservoir Cap. (mL)
Overall Height (mm)
Catalog Number
250
450
500
470
1,000
490
Back to top
Büchi condensers
Description/Model
Standard
Catalog Number
Plastic-coated
Catalog Number
A. Diagonal condenser A - with complete internal and external feed tubes, stopcock, 3 hose connectors, and vacuum seal
R114/R124
R200/R205
B. Vertical condenser V - with complete internal and external feed tubes, stopcock, 2 hose connectors, and vacuum seal
R114/R124
R200/R205
C. Cold-trap condenser C - with complete internal and external feed tubes, 2 stopcocks, 2 hose connectors, and vacuum seal
R114/R124
-
R200/R205
-
D. Vertical condenser S - with 2 hose connectors
R114/R124
-
R200/R205
E. Descending condenser E - with 2 hose connectors
R114/R124
-
R200/R205