Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Glassware Catalog
Extraction Thimbles - Sigma-Aldrich Glassware Catalog
 Miscellaneous  •  Thimbles  •  Systems
On this page:
Whatman cellulose extraction thimblesGlass microfiber thimbles
S&S cellulose extraction thimbles
For the separation of soluble substances from a mixture of solids by solvent extraction.
Whatman cellulose extraction thimbles
Pure cotton cellulose thimbles for extraction of environmental, food industry, medical, and other samples. Wall thickness: 1mm.
I.d. x H (mm)
Catalog Number
I.d. x H (mm)
Catalog Number
10 x 50
30 x 77**
22 x 65
30 x 80*,**
22 x 80*
30 x 100**
25 x 80
33 x 80*
25 x 90
33 x 94
25 x 100
33 x 118
26 x 60*
60 x 180
* Use with Soxhlet extraction units
** Use with Büchi extraction units
Back to top
Glass microfiber thimbles
High-purity borosilicate glass fiber thimbles for high temperature extractions up to 550°C and extractions for solvents incompatible with cellulose thimbles.
I.d. x H (mm)
Catalog Number
19 x 901*
30 x 100*
*Use with Soxhlet extraction units
Back to top
S&S cellulose extraction thimbles
Made from pure cotton linters. (S&S No. 603)
I.d. (mm)
Inside L (mm)
Catalog Number
10
50
19
90
25
80
26
60
33
80
33
94
43
123
Back to top