Glassware Catalog
Flat-Bottom Flasks - Sigma-Aldrich Glassware Catalog
Filtration
 •  Flat-Bottom  •  Round-Bottom  •  Multi-Neck  •  Schlenk  •  Septum-Inlet  •  Accessories
On this page:
Flat-bottom flasksCombustion apparatus
Flat-bottom flasks
Short neck, S.T. ground-glass joint.
Cap. (mL)
S.T. Joint
Catalog Number
25
14/20
50
14/20
50
24/40
100
14/20
100
24/40
250
14/20
250
24/40
500
14/20
500
24/40
1,000
24/40
2,000
24/40
Back to top
Combustion apparatus
1,000mL heavy-wall borosilicate glass conical flask with 35/25 ball joint stopper. Detachable platinum sample carrier is suspended from stopper hook. Pinch clamp holds stopper in place during combustion. Includes flask, stopper, platinum carrier, and pinch clamp. Order sample wrappers separately.
Description
Catalog Number
Apparatus
Black sample wrappers, 100 per package
Replacement parts
Flask, 1,000mL
Stopper
Pinch clamp
Sample carrier, Pt
Back to top