Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Glassware Catalog
Multi-Neck Flasks - Sigma-Aldrich Glassware Catalog
Filtration  •  Flat-Bottom  •  Round-Bottom  •  Multi-Neck  •  Schlenk  •  Septum-Inlet  •  Accessories
On this page:
Aldrich round-bottom distilling flasksJacketed 3-neck flasks
Three-neck round-bottom flasksSolid-phase peptide synthesis flasks
Four-neck round-bottom flasks
Five-neck round-bottom flasks
Aldrich round-bottom distilling flasks
Two-neck, angled, S.T. joints
Cap. (mL)
S.T. Joints
Catalog Number
50
14/20
100
14/20
100
24/40
250
14/20
250
24/40
250
29/32
500
14/20
500
24/40
500
29/32
500
34/45
(24/40 side)
1000
24/40
1000
29/32
1000
34/45
(24/40 side)
Three-neck, S.T. joints.
Cap. (mL)
S.T.
Joints
Angled
Catalog Number
Vertical
Catalog Number
50
14/20
100
14/20
100
24/40
250
14/20
250
24/40
500
14/20
500
24/40
500
29/32
500
34/45
(24/40 side)
1000
24/40
1000
29/32
1000
34/45
(24/40 side)
Back to top
Jacketed 3-neck flasks
Features:
  • Eliminates dead space for complete stirring of flask contents
  • Allows for complete draining without breaking down flask set-up
  • Liquid cooled or heated with liquid or steam
  • S.T. 24/40 side joints
Cap. (mL)
Center neck
S.T. joint
Half jacket
Catalog Number
Full jacket
Catalog Number
500
24/40
500
29/42
1,000
24/40
1,000
29/42
2,000
29/42
3,000
34/45
5,000
45/50
12,000
45/50
22,000
55/50
Back to top
Three-neck round-bottom flasks
All three necks are vertical in position. Additional flask and joint sizes
are available.
Cap. (mL)
Center neck
S.T. joint
Side neck
S.T. joints
Full jacket
Catalog Number
12,000
34/45
24/40
12,000
45/50
24/40
22,000
45/50
24/40
22,000
45/50
45/50
22,000
45/50
65/40 (socket)
50,000
45/50
24/40
Back to top
Solid-phase peptide synthesis flasks
Has three S.T. 24/40 side joints and a center joint that varies in size according to capacity of flask. Coarse frit (25 to 505m) and PTFE Rotaflo stopcock permit rapid filtration and washing of resin.
Cap. (mL)
Center
S.T. Joint
Catalog Number
1,000
24/40
5,000
34/45
12,000
45/50
Back to top
Four-neck round-bottom flasks
All four necks are vertical in position. The fourth neck is set at a 90° angle to the three vertical in-line necks. Additional flask and joint sizes are available.
Cap.
(mL)
Center neck
S.T. joint
Side neck
S.T. joints
90° Angle neck S.T.joint
Catalog Number
12,000
34/45
24/40
24/40
12,000
45/50
24/40
24/40
12,000
55/50
24/40
24/40
12,000
55/50
65/40
(socket)
24/40
12,000
75/50
(socket)
65/40
(socket)
24/40
12,000
75/50
(socket)
75/50
(socket)
24/40
22,000
45/50
24/40
24/40
22,000
45/50
65/40
(socket)
24/40
22,000
55/50
65/40
(socket)
24/40
22,000
75/50
(socket)
75/50
(socket)
24/40
50,000
45/50
24/40
24/40
50,000
102/75
(socket)
75/50
(socket)
24/40
72,000
102/75
(socket)
75/50
(socket)
24/40
Back to top
Five-neck round-bottom flasks
All five necks are vertical in position. The flask is configured with a center neck, two side necks and two front necks. Additional flask and joint sizes are available.
Cap.
(mL)
Center neck
S.T. joint
Side neck
S.T. joints
Front neck
S.T. joints
Catalog Number
12,000
45/50
24/40
24/40
22,000
45/50
24/40
24/40
22,000
45/50
65/40
(socket)
24/40
50,000
45/50
24/40
24/40
50,000
55/50
65/40
(socket)
24/40
50,000
102/75 (socket)
75/50
(socket)
24/40
72,000
102/75 (socket)
75/50
(socket)
24/40
Back to top
See the Heating Mantles Section for the
full listings of Glas-Col heating mantles.