Glassware Catalog
Septum-Inlet Flasks - Sigma-Aldrich Glassware Catalog
Filtration  •  Flat-Bottom  •  Round-Bottom  •  Multi-Neck  •  Schlenk  •  Septum-Inlet  •  Accessories
On this page:
Round-bottom flasks with septum-inlet
Storage flasks with stopcock-equipped septum-inlet
Round-bottom flasks with septum-inlet
With ground-glass joint. Use septum Z10,072-2 or Z12,435-4.
Flask size (mL)
S.T joint
Catalog Number
25
14/20
50
14/20
100
14/20
100
19/22
100
24/40
250
14/20
250
19/22
250
24/40
250
29/32
500
24/40
500
29/32
Back to top
Storage flasks with stopcock-equipped septum-inlet
With ground-glass joint. Use septum Z10,072-2 or Z12,435-4.
Flask size (mL)
S.T joint
Catalog Number
2mm glass stopcock
50
14/20
100
14/20
250
24/40
250
29/32
4mm glass stopcock
500
24/40
500
29/32
1000
24/40
1000
29/32
2mm PTFE stopcock
50
14/20
100
14/20
100
19/22
250
14/20
250
19/22
250
24/40
250
29/32
4mm PTFE stopcock
500
24/40
500
29/32
1,000
24/40
1,000
29/32
Back to top