Glassware Catalog
Buchner Funnels - Sigma-Aldrich Glassware Catalog
Addition  •  Buchner  •  Filter/Drying  •  Miscellaneous
On this page: Aldrich constant-temperature Buchner funnel
Aldrich vacuum-jacketed Buchner funnelAce Buchner funnels
Aldrich constant-temperature Buchner funnel
With jacket for circulating steam or coolant. 60mL capacity, 180mm H, 40mm diam. frit, 25 to 50 µm porosity. Drip-tip joint and vacuum take-off.
S.T. joint Catalog Number
24/40 Z17,583-8
29/32 Z51,775-5
Back to top
Aldrich vacuum-jacketed Buchner funnel
Silvered, 60mL capacity, 180 mm H, with 40mm diam. frit, 25 to 50 µm porosity. Drip-tip joint and vacuum take-off.
S.T. joint Catalog Number
24/40 Z17,581-1
29/32 Z51,776-3
Back to top
Ace Buchner funnels
With glass frit, hose connection, and S.T. 24/40 joint.
Porosity (µm)
Catalog Number
Disc diam.: 40mm; capacity: 60mL
145 - 175
70 - 100
25 - 50
10 - 20
4 - 8
Disc diam.: 65mm; capacity: 140mL
145 - 175
70 - 100
25 - 50
10 - 20
4 - 8
Disc diam.: 80mm; capacity: 350mL
145 - 175
70 - 100
25 - 50
10 - 20
4 - 8
Back to top