Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Glassware Catalog
Addition Funnels - Sigma-Aldrich Glassware Catalog
Addition  •  Buchner  •  Filter/Drying  •  Miscellaneous
On this page:
Addition funnels (2mm PTFE stopcock)Addition funnels with pressure-equalization arm
Addition funnels with PTFE needle valveLarge capacity addition funnels
Addition funnels
(T-bore stopcock)
Addition funnels
Cylindrical style with 2mm PTFE stopcock (4mm for 500mL capacity).
Cap. (mL)
Overall H (mm)
S.T. joints
Catalog Number
10
170
14/20
25
220
14/20
50
280
14/20
100
300
14/20
100
320
24/40
100
320
29/32
250
370
24/40
250
370
29/32
500
430
24/40
500
430
29/32
Back to top
Addition funnels with pressure-equalization arm
Cylindrical style with 2mm PTFE stopcock (4mm for 500mL capacity).
Cap. (mL)
Overall H (mm)
S.T. joints
Catalog Number
10
170
14/20
25
220
14/20
50
280
14/20
100
300
14/20
100
320
24/40
100
320
29/32
250
370
24/40
250
370
29/32
500
430
24/40
500
430
29/32
Back to top
Addition funnels with PTFE needle valve
With pressure-equalization arm, 0-3mm PTFE needle valve for precise addition-rate control.
Cap.
(mL)
Overall H
(mm)
S.T.
Joints
Jacketed
Catalog Number
Unjacketed
Catalog Number
100
430
24/40
100
430
29/32
250
500
24/40
250
500
29/32
500
550
24/40
500
550
29/32
Back to top
Large capacity addition funnels
With pressure-equalization arm, 0-3mm PTFE needle stopcock for precise addition-rate control.
Cap.
(mL)
O.d.
(mm)
Overall H (mm)
S.T. 24/40 joints
Catalog Number
S.T. 29/32 joints
Catalog Number
1,000
180
560
2,000
100
580
Back to top
Addition funnels
With 4mm T-bore glass stopcock on pressure-equalization arm and 0-3mm PTFE needle valve.
Cap.
(mL)
O.d.
(mm)
Overall H (mm)
S.T. 24/40 joints
Catalog Number
S.T. 29/32 joints
Catalog Number
1,000
180
560
2,000
100
580
Back to top