Labware > Glassware Center > Glassware Kits & Components > Student Lab Glassware Kits
Glassware Catalog
Student Lab Glassware Kits
Glassware with standard 14/20 ground glass joints provides a high quality lowcost solution for school laboratories and research. Customize the basic 14/20 kit with your choice of glass components and accessories.
 Z122548Lblue Tube, drying Adapter, vacuum Adapter, vacuum-distilling Flask, round-bottom:100 mL Adapter, connecting, 3-Way Flask, round bottom, 100 mL 3-neck, angled Column, Vigreux distilling Adapter, Claisen Stopper, hex-head, hollow Adapter, gas-inlet/thermometer Tube, gas-inlet Funnel, addition
            50 mL with PTFE stopcock Flask, round-bottom:25 mL Condenser, Liebig Flask, round-bottom: 50 mL
Kit
Quantity
Description Product
Number
1 StudentLabs (14/20 joints) Kit Z122548
1 Adapter, Claisen Z413429
1 Adapter connecting 3-way Z122408
1 Adapter glass-inlet/thermometer Z543748
1 Adapter, vacuum Z122394
1 Adapter, vacuum-distilling Z122386
1 Column, Vigreux distilling Z122475
1 Condenser Liebig Z122416
1 Flask, round-bottom: 25mL Z100641
1 Flask, round-bottom: 50 mL Z122432
1 Flask, round-bottom: 100 mL Z100676
1 Flask, round-bottom 100mL
3-neck angled
Z122459
1 Funnel addition 50mL
with PTFE stopcock
Z122467
2 Stopper hex head hollow Z122483
1 Tube drying Z122491
1 Tube gas-inlet Z122513
1 Foamlined storage case
(Not shown)
Click on the Glassware piece or description to link to a larger view
Student Labs Component Tour (PDF 136KB)
Additional Components for the Student Lab  14/20 joints Kit
Return to the Aldrich Lab Kits & Components page
Return to the Aldrich Glassware home page