Labware
Glas-Col Series TM Heating Mantles - Sigma-Aldrich Glassware Catalog
Glas-Col Series O  •  Glas-Col Series TM  •  Glas-Col Controllers
On this page:
Hemispherical heating mantlesReaction flask mantlesBuchner funnel mantles
Glas-Col series STM heating mantlesLarge capacity StirMantles
Series TM mantles have an aluminum exterior for extreme durability.
  • Interior made from flexible fabrics
  • Maximum 450°C internal operating temperature
  • CSA compliant
All mantles include a 4ft, 3-wire cord and locking connector. A temperature controller is required for operation.
Hemispherical heating mantles
The mantle covers the bottom half of single and multi-neck flask allowing for full view of contents while heating.
Flask
size (mL)
Watts
115V
Catalog Number
50
160
100
180
125
180
200
100
250
180
300
180
500
270
1,000
380
2,000
500
3,000
500
5,000
600
Back to top
Reaction flask mantles
Provides even heat for reaction kettlesideal for resinous materials. Order heating mantle corresponding to flask manufacturer and size.
Flask
size (mL)
Watts
(Ace/Corning)
Ace
Catalog Number
Corning
Catalog Number
500
-/270
-
1,000
300/335
2,000
450/470
3,000
600/550
4,000
-/750
-
Back to top
Buchner funnel mantles
Provides convenient support for funnel and gentle heat for solutions that must be kept warm during transfer. Durable support bracket slides over mast of ring stand, enabling mantle to be positioned at any convenient height above receiver. Accepts rods up to 1/2in. diam.
  • Detachable 4ft, 3-wire cord and locking connector
  • 115V units are CSA compliant
Mantles for fixed plate Buchner funnels
Top diam. (mL)
Filter size (mm)
Coors No.
115V
Catalog Number
230V
Catalog Number
2.75
55
60240
-
3.50
70
60242
-
4.25
90
60243
-
4.94
110
60244
-
5.63
124
60245
6.75
150
60246
8.38
185
60247
10.63
240
60248
Mantles for fritted disc Buchner funnels (Corning 36060 or Kimble 28400)
Cap (mL)
Disc. diam. (mm)
115V
Catalog Number
230V
Catalog Number
150
70
350
90
600
100
2,000
140
3,000
165
Back to top
Glas-Col series STM heating mantles
The Series STM mantles perform the same function as Series TM heating mantles, plus operations requiring more heat. 12 and 22L mantles have 3in. legs for cool lab benches, are sealed, and have inert gas purge fittings. A temperature controller is required for operation.
Caution: Series STM mantles are recommended for use with flammable solvents only when used with a Poncho safety shield. Please contact our Technical Service department for information.
  • Maximum 650°C internal operating temperature
  • 115V units are CSA compliant (except 100 to 300mL units)
  • Detachable 4ft, 3-wire cord and locking connector (except 12 and 22L models which have a terminal box)
Flask
size (mL)
Watts
115V
Catalog Number
230V
Catalog Number
100
230
125
230
200
300
250
470
300
470
500
470
1,000
600
2,000
870
3,000
970
5,000
2 @ 700
12,000
2 @ 1,000
22,000
3 @ 1,000
Back to top
Large capacity StirMantles
Has the power to mix volumes up to 72L with a magnetic stir bar, eliminating the need for an overhead stirrer. Ideal for continuous distillations. Supporting feet allow bench clearance, and maintain even air flow. Order a temperature controller separately.
  • Stirring range: 100 to 1,850rpm
  • Variable-speed stir control
  • 4ft detachable cord (except on 50 and 72L which have terminal boxes)
  • Not CE compliant
Flask
size (L)
Watts
115V
Catalog Number
230V
Catalog Number
12
2 x 650
22
2 x 770
50
3 x 1,000
72
2 x 2,000