Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Glassware Catalog
Glas-Col Series O Heating Mantles - Sigma-Aldrich Glassware Catalog
Glas-Col Series O  •  Glas-Col Series TM  •  Glas-Col Controllers
On this page:
Hemispherical heating mantlesBuchner funnel mantles
Series O mantles are the original and the most popular heating mantles.
  • Made from flexible fabrics
  • Fit easily into tight spaces
  • Maximum 450°C internal operating temperature
  • CSA and CE mark of conformity
All mantles include a 4ft, 2-wire cord and locking connector. A temperature controller is required for operation.
Hemispherical heating mantles
The mantles cover the bottom half of single and multi-neck flasks, allowing for full view of contents.
Flask size (mL)
Watts
115V
Catalog Number
230V
Catalog Number
50
60
-
100
80
-
125
80
-
200
100
-
250
180
300
180
-
500
270
1,000
380
2,000
500
3,000
500
-
5,000
600
12,000
2 @ 650
-
Back to top
Buchner funnel mantles
  • Provides gentle heat for solutions that must be kept warm during transfer
  • Not CE compliant
Top diam. (in.)
Coors
Funnel No.
Watts
115V
Catalog Number
230V
Catalog Number
12.56
60240
50
-
13.38
60242
50
-
14.06
60243
70
-
14.81
60244
70
-
15.38
60245
100
16.50
60246
220
18.13
60247
300
10.5
60248
350
Back to top