|
Glassware Catalog
Kugelrohr Apparatus
|
Diazomethane • Kugelrohr • Miscellaneous
Kugelrohr Apparatus • Accessories
On this page: Ball-Tube • Short-Path
 |
Kugelrohr apparatus shown with KNF Laboport vacuum pump (sold separately).
- Distills heat-sensitive compounds
- Distills liquids and low-melting solids from polymers and tars
- Distills with minimal holdup and sample loss
- Removes color and particulates
- Can be used for sublimation/solvent evaporation
|
• Distills the Most Difficult Materials •
|
Step 1
|
Oven flask is filled one-third full with distillable material, placed carefully in oven, and connected to the horizontal receiving flasks outside the oven. |
|
Step 2
|
Connect the glassware to the rotary drive and the drive to a vacuum pump.* Switch the vacuum drive on to turn the distillation train 360° to speed distillation, ensure even heating, and prevent bumping. Start the vacuum pump.
*Use of a vacuum trap between the rotary drive and the pump is recommended to protect the pump. See the Equipment Section of the Aldrich Handbook for vacuum traps. |
|
Step 3
|
When the correct vacuum is attained, set the distillation temperature on the air-bath oven and begin distillation. The digital temperature controller displays both "set" and "actual" air-bath temperature. |
|
Step 4
|
Collect distillate in the horizontal ball-tubes outside of oven. An ice-filled polypropylene cooling tray quickly condenses distillate in receiving ball-tubes. |
Specifications:
- Micro to macro distillation capability
- Accommodates flask sizes 10mL to 2L
- Air-bath oven
- SS wall with leakproof seal at the base contains spills
- Grounded heating element prevents electrical shock
- Detachable power cord, on/off switch, adjustable rubber feet, and interchangeable PTFE bearing set for flasks with S. T.14/20 and S. T. 24/40 joints
- Distill under vacuum to 0.05mm Hg
- Single-speed rotary drive is optimized for Kugelrohr distillations
- Detachable power cord, on/off switch, adjustable rubber feet, built-in stabilizing clamp
- Automatic temperature controller
- Maintains oven temperature up to 220°C, ±1°C
- Type-K thermocouple ensures fast, accurate temperature measurements inside of oven
- Automatically turns off power to oven if thermocouple fails or disconnects
- CE Compliant
|
Includes the following components:
- air-bath oven with digital temperature controller
- glassware set consisting of a straight tube with hose connection
- 25 and 100mL round-bottom oven flasks
- 25 and 100mL single bulb ball-tube flasks with S. T. 14/20 joints
- rotary drive.
|
Volts
|
Catalog Number
|
|
115
|
|
|
230