|
Glassware Catalog
Kugelrohr Accessories
|
Diazomethane • Kugelrohr • Miscellaneous
Kugelrohr Apparatus • Accessories
On this page: Oven flasks • Single-bulb ball-tubes • Dual-bulb ball-tubes • Straight tubes • Closing cap Standard Kugelrohr glassware set • Large-capacity Kugelrohr glassware sets
Oven flasks
 |
Heavy-wall borosilicate glass with ground-glass joints.
|
Cap. (mL)
|
Catalog Number
|
|
With S.T. 14/20 joint
|
|
10
|
|
|
25
|
|
|
50
|
|
|
100
|
|
|
250
|
|
|
500
|
|
| |
|
With S.T. 24/40 joint
|
|
1,000
|
|
|
Back to top
Single-bulb ball-tubes
 |
Arranged horizontally outside of Kugelrohr oven, the ball-tubes cool and collect distillate with minimal loss of material.
|
Cap. (mL)
|
Plain Catalog Number
|
w/ Drip-tip Catalog Number
|
| With S.T. 14/20 joints |
|
10
|
25
|
50
|
100
|
250
|
500
|
|
|
| |
| With S.T. 24/40 joints |
|
250
|
500
|
1,000
|
2,000
|
4,000
|
Back to top
Dual-bulb ball-tubes
 |
S.T. joint on one end to connect to oven flask and a hose connector on the other for connection to the motor drive unit.
|
Cap. (mL)
|
S.T. Joint
|
Catalog Number
|
|
25
|
14/20
|
|
|
50
|
14/20
|
|
|
100
|
14/20
|
|
|
250
|
24/40
|
|
|
500
|
24/40
|
|
|
Back to top
Straight tubes
 |
Provides a S.T. joint connection to single bulb ball-tube flasks and a hose connector to motor drive.
|
S.T. Joint
|
Catalog Number
|
|
14/20
|
|
|
24/40
|
|
|
Back to top
Closing cap
 |
Female S.T. joint closes end of ball-tube to prevent spillage of distillate during handling.
|
S.T. Joint
|
Catalog Number
|
|
14/20F
|
|
|
Back to top
Standard Kugelrohr glassware set
|
|
Set includes a straight tube with hose connection, 25 and 100mL round-bottom oven flasks, and 25 and 100mL single bulb receiver flasks with S.T. 14/20 joints.
Z100501 |
Back to top
Large-capacity Kugelrohr glassware sets with System 45 compatible connections 
Sets contain: 1 Straight tube with removable 1/4in. i.d. hose connection 1 Single-bulb ball-tube flask 1 Round-bottom oven flask 2 PBT/PTFE screw-thread connectors 3 Solid-top caps, PBT with PTFE liner
PBT/PTFE connectors resist heat to 200°C and may be used interchangeably with 1- and 2-liter glassware. Special "reverse threading" on these flasks prevents connections from loosening during rotation.
|
Description
|
Catalog Number
|
|
1-Liter glassware set
|
|
|
2-Liter glassware set