Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Glassware Catalog
Lattice Systems - Sigma-Aldrich Glassware Catalog
On this page:
Aldrich Lattice System SetsAldrich lattice rodsMounting adaptersChannel clampLattice-rod clamps"T"-handle screwsQuick-action lattice-rod clampsBase platesSliding-extension-clamp couplingsExtension coupling
Aldrich Custom Lattice Systems
  • Pre-cut lattice-rods and hardware are available from stock for immediate shipment
  • Custom-length lattice-rods can be ordered from Aldrich
  • Special pricing for bulk orders of rods and hardware
Please contact your local Sigma-Aldrich Location for a quotation on custom-length lattice rods or bulk orders.
Back to top
Lattice System Hardware
Aldrich lattice rods
Rods are 1/2in. diameter.
L (in.)
Fiberglass
Catalog Number
Aluminum
Catalog Number
316 SS
Catalog Number
8
-
-
12
-
-
24
30
-
-
34
-
-
44
-
-
48
72
96
-
108
-
-
144
-
-
Back to top
All hardware is for 1/2in. diameter lattice rods.
Mounting adapters
For mounting lattice-support rods to benchtops already equipped with 1/2in. countersunk fittings. Aluminum with SS set screws 1/2in. NPTM thread.
Z12,498-2
Back to top
Channel clamp
For fastening lattice-support rods to the channel frames in Aldrich lattice systems above. Can be loosened from frame to position rods.
Z12,516-4
Back to top
Lattice-rod clamps
Back to top
Die-cast aluminum alloy clamp equipped with Allen set screws. Assembles without dismantling the existing lattice-rod setup.
Z10,717-4
"T"-handle screws
Back to top
Converts lattice-rod clamp (Z10,717-4) to quick-action clamp (Z10,858-8).
Z10,859-6
Quick-action lattice-rod clamps
Back to top
Similar to above, but equipped with "T"-handle and SS screws, permitting rod adjustment without tools.
Z10,858-8
Base plates
Back to top
Mounts lattice-support rods to benchtop, wall, or ceiling. Unthreaded type provides perpendicular support. Swivel type permits locking a rod in any position up to 22.5° from vertical using two Allen screws set at right angles to each other.
Description
Catalog Number
Unthreaded type
Swivel type
Sliding-extension-clamp couplings
Back to top
38mm L, with thumbscrews. Connects lattice rods
side-by-side for length adjustments.
Z19,325-9
Extension coupling
Back to top
76mm L, with thumbscrews. Connects lattice rods end-to-end.
Z19,324-0