|
Glassware Catalog
Miscellaneous Distillation Products
|
Diazomethane • Kugelrohr • Miscellaneous
On this page: Büchi Model B580 GKR oven for distillation • Micro short-path distillation apparatus • Short-path distillation apparatus • Solvent-still-head apparatus • Solvent still heads • Azeotropic distillation apparatus • Dropping funnel
Büchi Model B580 GKR oven for distillation
 |
The small size flasks in this ball-tube oven make it an ideal system for high temperature distillation of small quantities of liquid, for separating solvents from non-volatile, oily components in a mixture or for evaporating quantities of sample that are too small for rotary evaporators. Includes 3 x 20mL ball-tube assembly, ball-tube drive unit and 60mL rotation drying flask.
|
- Variable rotation speed, 0 to 50rpm
- Cooling device cold trap can be cooled down to -80°C
- Several different inserts, ball-tubes with 2, 3 or 4 balls are available
- External ball-tube drive unit for constant rotation
- Linear drying tube holds up to 60mL of solids
- CE approved
|
Volts
|
Catalog Number
|
|
115
|
|
|
230
|
|
Back to top
Micro short-path distillation apparatus
 |
Threaded S.T. 14/20 joints.
|
Description
|
Catalog Number
|
| Complete apparatus |
|
| Standard head |
|
| Jacketed head |
|
Replacement parts |
|
| Standard head |
|
| Jacketed head |
|
| Cow receiver |
|
| Recovery flask, 5mL |
|
| Distillation flask, 10mL |
|
| Gas-inlet tube |
|
| Thermometer, -10 to 250°C, 40mm immersion L |
|
|
Back to top
Short-path distillation apparatus
 |
Threaded S.T. 24/40 joints. 100mL two-neck flask and 50mL recovery flask.
|
Description
|
Catalog Number
|
| Complete apparatus |
|
| Standard head |
|
| Jacketed head |
|
Replacement parts |
|
| Standard head |
|
| Jacketed head |
|
| Receiver flask, 50mL |
|
| Two-neck flask, 100mL |
|
| Two-neck flask, 250mL |
|
| Gas-inlet adapter |
|
| Gas-inlet tube |
|
Thermometer, -10 to 260°C, 76mm immersion L |
|
|
Back to top
Solvent-still-head apparatus
 |
S.T. 24/40 joints. Includes still head, column with Raschig rings, Friedrichs condenser with water-inlet tube, one-neck round-bottom flask, stopper, and nitrogen-inlet adapter. Designed by Paul F. Hudrlik- may be used for continuous distillation.
|
Cap. (mL)
|
RB flask size (mL)
|
Catalog Number
|
|
250*
|
2,000
|
|
|
250**
|
2,000
|
|
|
500
|
5,000
|
|
|
1,000
|
5,000
|
|
| |
| Replacement still heads |
|
250*
|
-
|
|
|
500
|
-
|
|
|
1,000
|
-
|
|
|
* Has S.T. 14/20 joint to receiver flask ** Has S.T. 29/32 joints and S.T. 14.5/23 joint to receiver flask
Back to top
Solvent still heads
|
|
With return finger and 4mm PTFE stopcock.
|
Cap.(mL)
|
S.T Joint
|
Catalog Number
|
|
250
|
14/20
|
|
|
500
|
24/40
|
|
|
1,000
|
24/40
|
|
|
Back to top
Azeotropic distillation apparatus
All connections are made using greaseless, S.T. 14/20 screw-thread joints. All parts are interchangeable with those contained in the Aldrich All-In-One Glassware kit, Z166898. Complete apparatus includes distillation head and 100mL round-bottom flask.
|
 |
 Azeotropic Technique:
- The solvent/H2O azeotrope is cooled completely by a water-cooled condenser and directed to the extraction reservoir (filled with 4Å molecular sieves through a S.T. 14/20 joint). The reservoir joint* can also be used as a septum inlet for the removal of water via syringe
- The solvent and any volatile reagents are recycled through the sieves at ambient temperature
- Additional reagent can be added to the reservoir (via dropping funnel connected at the reservoir joint) as a means of slow addition to a mixture at reflux
Z250325
*Use with rubber septa Z100749, Z124370, Z167282, Z124591
1. Barta, N.S.; Paulvannan, K.; Schwarz, J.B.; Stille, J.R. Synth. Commun. 1994, 24, 583. |
Back to top
Dropping funnel
50mL, with PTFE stopcock and S.T. 14/20 screw-thread joints. Z167541