On this page: Aldrich 5mm NMR tube filling manifolds • Aldrich 5mm NMR tube evacuation manifolds 5mm NMR tube with PTFE valve • NMR-tube Dewar flask • Pocket NMR tube carriers
Aldrich 5mm NMR tube filling manifolds
NMR Tube Filling Technique
- Insert NMR tube through the open top cap and Viton O-ring (see figure 1)
- Evacuate NMR tube
- Fill NMR tube with a syringe through the PTFE-faced silicone septa (see figure 2)
- Tip-off NMR tube or remove tube and cap (see figure 3)
|
 |
Manifolds have 0-10mm bore PTFE valves. The multiple filling station units allow removal of one NMR tube while the remaining tube(s) stay under vacuum.
|
Filling Stations
|
L (mm)
|
Catalog Number
|
|
1
|
100
|
|
|
2
|
150
|
|
|
5
|
300
|
|
|
Replacement Parts
| Description |
Catalog Number
|
| Replacement PTFE-faced silicone septa |
|
| Replacement screw caps |
|
| Replacement Viton O-rings |
|
|
Accessories
|
|
|
| For vacuum-thaw applications, use the Aldrich NMR tube Dewar flask. |
When the tip-off technique is used, the MT-51 butane torch (listed below) generates a 1,300°C pinpoint flame. |
| Description
|
Catalog Number
|
| NMR tube Dewar flask |
|
| MT-51 handheld torch |
|
| (Torch shipped empty, order butane below) |
| Butane torch fuel, 26g of butane fuel per can |
|
Back to topAldrich 5mm NMR tube evacuation manifolds   Manifolds have 0-10mm bore PTFE valves. The multiple evacuation station units allow removal of one NMR tube while the remaining tube(s) stay under vacuum.
|
Evacuation stations
|
L (mm)
|
Catalog Number
|
|
1
|
100
|
|
|
2
|
150
|
|
|
5
|
300
|
|
Back to top Accessories5mm NMR tube with PTFE valve Improve the handiling of NMR samples. A convenient alternative to flame-sealing.
Z18,397-0Back to top
NMR-tube Dewar flask Compact size minimizes volume of liquid nitrogen required. Borosilicate glass with protective aluminum housing. 2.0 i.d. x 23cm depth.
Z24,729-4
Back to topPocket NMR tube carriers
 |
Unbreakable NMR tube carrier fits into lab-coat pocket like a pen. Has a snap-on cap and a shirt clip that secures carrier in pocket. Holds one 5mm diam. x 7in. L NMR tube.
|
Color
|
Catalog Number
|
| Red |
|
| White |
|
| Blue |
|
| Color assortment pack |
|
| One of each color carrier; red, white and blue |