Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Labware > Glassware Center > Specialty NMR Tube Cleaners
Glassware Catalog
Specialty NMR Tube Cleaners
Aldrich NMR Tube Cleaners  •  Specialty NMR Tube Cleaners  •  NMR Tube Manifolds
On this page:
Dual-action NMR tube cleanersHigh capacity NMR tube cleaner
NMR tube cleaner with wash and dry cycleFive-position NMR tube cleaner system
Dual-action NMR tube cleaners
al_Z41417L
  • For high volume cleaning, no o-rings needed
  • Specially designed to prevent accidental breakage of tubes
  • Highly efficient solvent jets improve cleaning and reduce solvent usage
  • For 7 or 8in. L, 5mm NMR tubes
Number of positions
S.T. Joint
Catalog Number
One
24/40
One
29/32
Three
24/40
Three
29/32
Reservoir flask, 1,000 mL
24/40
29/32
Back to top
High capacity NMR tube cleaner
al_Z23921LSingle position tube cleaner with 100mL solvent reservoir for less frequent filling.
S.T. Joint
For Tube (mm)
Catalog Number
24/40
5
29/32
5
Back to top
NMR tube cleaner with wash and dry cycle
al_Z23925La al_Z23925Lb
Rotating the 2mm PTFE stopcock 1/4 turn controls the washing and drying cycles. Integral check-valve prevents back flow of spent solvent into reservoir.
al_glassware_note96
S.T. Joint
For Tube (mm)
Catalog Number
Tooled end
5
24/40
5
29/32
5
Back to top
Five-position NMR tube cleaner system
al_Z42284L
Washes up to five 5mm, 7 or 8in. L NMR tubes at once (Figure 1). NMR tube caps can be used to plug holes if less than five tubes are to be cleaned. Cleaning solvent is pulled from an external container via PTFE tube, eliminating repetitious filling of side mounted reservoirs. Note: Bottle for cleaning solvent is not included.
  • Totally inert, cleaning solvents contact borosilicate glass and PTFE
  • No ground joints to freeze, eliminates grease
  • Top PTFE tube holder provides a vacuum-tight seal
  • Modular components are easy to replace
Simple to use:
  1. Slide O-rings approx. 25mm down on NMR tubes
  2. Insert NMR tubes into top adapter
  3. Turn on vacuum, make sure all O-rings are seated in top adapter
  4. Open solvent valve to wash tubes
  5. Turn solvent valve 90° to air dry tubes
Z42,284-3
Back to top