On this page: Dual-action NMR tube cleaners • High capacity NMR tube cleaner NMR tube cleaner with wash and dry cycle • Five-position NMR tube cleaner system
Dual-action NMR tube cleaners
|
- For high volume cleaning, no o-rings needed
- Specially designed to prevent accidental breakage of tubes
- Highly efficient solvent jets improve cleaning and reduce solvent usage
- For 7 or 8in. L, 5mm NMR tubes
|
Number of positions
|
S.T. Joint
|
Catalog Number
|
|
One
|
24/40
|
|
|
One
|
29/32
|
|
|
Three
|
24/40
|
|
|
Three
|
29/32
|
|
| |
| Reservoir flask, 1,000 mL |
24/40
|
29/32
|
|
|
Back to top
High capacity NMR tube cleaner
Single position tube cleaner with 100mL solvent reservoir for less frequent filling.
|
S.T. Joint
|
For Tube (mm)
|
Catalog Number
|
|
24/40
|
5
|
|
|
29/32
|
5
|
|
Back to top
NMR tube cleaner with wash and dry cycle
| Rotating the 2mm PTFE stopcock 1/4 turn controls the washing and drying cycles. Integral check-valve prevents back flow of spent solvent into reservoir. |
|
|
S.T. Joint
|
For Tube (mm)
|
Catalog Number
|
|
Tooled end
|
5
|
|
|
24/40
|
5
|
|
|
29/32
|
5
|
|
Back to top
Five-position NMR tube cleaner system
Washes up to five 5mm, 7 or 8in. L NMR tubes at once (Figure 1). NMR tube caps can be used to plug holes if less than five tubes are to be cleaned. Cleaning solvent is pulled from an external container via PTFE tube, eliminating repetitious filling of side mounted reservoirs. Note: Bottle for cleaning solvent is not included.
- Totally inert, cleaning solvents contact borosilicate glass and PTFE
- No ground joints to freeze, eliminates grease
- Top PTFE tube holder provides a vacuum-tight seal
- Modular components are easy to replace
Simple to use:
- Slide O-rings approx. 25mm down on NMR tubes
- Insert NMR tubes into top adapter
- Turn on vacuum, make sure all O-rings are seated in top adapter
- Open solvent valve to wash tubes
- Turn solvent valve 90° to air dry tubes
Z42,284-3
Back to top |