INTEGRAL SOLVENT RESERVOIR (Figure 1)
Models A, B, C
A PP tube cap placed on the closed end of an inverted NMR tube holds the tube in place and seals the system to allow efficient vacuum. Applied suction sprays solvent inside and against the closed end of tube for thorough cleaning.
Model D
As above, except the ground-glass stopper is the cap.
|
Model
|
S.T. Joint
|
For tube (mm)
|
Catalog Number
|
|
A
|
Tooled end
|
5
|
|
|
B
|
24/40
|
5
|
29/32
|
5
|
24/40
|
10
|
|
|
C
|
24/40
|
5
|
29/32
|
5
|
24/40
|
10
|
|
|
D
|
24/40
|
5
|
|
Reservoir flask (1,000mL) with hose barb |
| |
24/40
|
|
|
| |
29/32