|
On this page: Vessels with side port and fritted discs • Vessels with cap and fritted disc Vessels with side port and removable fritted disc (GL 18 threaded side port) Threaded vessel adapters • Vessels with removable fritted disc Vessels with side port and removable fritted disc (S.T. 14/20 outer joint side port) Peptide vessels • Jacketed vessels
Vessels with side port and fritted discs
Cylindrical vessel has a GL 18 threaded side port and sealed top and bottom, coarse porosity, fritted discs located to minimize dead space. Top and bottom PTFE plug-style valves reduce the potential for breakage associated with glass stopcocks. 2mm PTFE valves require 1/4-turn to open or close. Valve stems have a 1/4-28 i.d. thread while the o.d. is sized for a 3/8in. compression fitting. Complete with two 2mm PTFE valves and side port cap. (Corning 46800)
|
Cap. (mL)
|
Distance between discs (mm)
|
Disc diam. (mm)
|
Catalog Number
|
|
20
|
60
|
20
|
|
|
60
|
105
|
25
|
|
|
125
|
110
|
30
|
|
|
250
|
160
|
40
|
|
|
500
|
210
|
50
|
|
Back to Top
Vessels with cap and fritted disc
Cylindrical vessel has a 32mm cap with PTFE-faced liner. Vessel comes with a sealed coarse porosity fritted disc located to minimize dead space. 2mm PTFE bottom valve reduces the potential for breakage associated with glass stopcocks. 2mm PTFE valve requires 1/4-turn to open or close. Valve stem has a 1/4-28 i.d. thread while the o.d. is sized for a 3/8 in. compression fitting. Complete with cap, liner, and 2mm bottom valve. (Corning 46802)
|
Cap. (mL)
|
Distance between discs (mm)
|
Disc diam. (mm)
|
Catalog Number
|
|
20
|
60
|
20
|
|
|
60
|
105
|
25
|
|
|
125
|
110
|
30
|
|
|
250
|
160
|
40
|
|
|
500
|
210
|
50
|
|
Back to Top
Vessels with side port and removable fritted disc 
Cylindrical style vessel has a 2mm PTFE valve at the top, with a GL 18 threaded side port. The 32mm threaded bottom valve assembly incorporates System 45 technology. The coarse fritted disc is easily removed from the PTFE adapter for cleaning and/or replacement. PTFE top and bottom valves require 1/4-turn to open or close. Valve stems have a 1/4-28 internal thread in the stem which allows a variety of adapters to be used. For male 1/4-28 and male Luer adapters, see Cat. No. Z41,741-6 and Z41,742-4 below. (Corning 46804)
|
Approx. cap. (mL)
|
L x diam. (mm)
|
Catalog Number
|
| Complete |
|
20
|
50 x 28
|
|
|
60
|
100 x 32
|
|
|
125
|
105 x 45
|
|
|
250
|
155 x 51
|
|
|
500
|
220 x 70
|
|
| Vessel only |
|
20
|
-
|
|
|
60
|
-
|
|
|
125
|
-
|
|
|
250
|
-
|
|
|
500
|
-
|
|
Back to Top
Threaded vessel adapters (Corning 42290)
|
Approx. cap. (mL)
|
L x diam. (mm)
|
Catalog Number
|
| Complete |
|
20
|
60
|
100 x 32
|
|
|
125
|
105 x 45
|
|
|
250
|
155 x 51
|
|
|
500
|
220 x 70
|
|
| Vessel only |
|
20
|
-
|
|
|
60
|
-
|
|
|
125
|
-
|
|
|
250
|
-
|
|
|
500
|
-
|
|
Back to Top
Vessels with side port and removable fritted disc
 |
Cylindrical style vessel has a 2mm PTFE valve at the top, with a S.T. 14/20 outer joint side port. The 32mm threaded bottom valve assembly incorporates System 45 technology. The coarse fritted disc is easily removed from the PTFE adapter for cleaning and/or replacement. PTFE bottom valve requires 1/4-turn to open or close. Valve stems have a 1/4-28 internal thread in the stem which allows a variety of adapters to be screwed into place. For male 1/4-28 and male Luer adapters, see Cat. No. Z41,741-6 and Z41,742-4 above. (Corning 46808)
|
Approx. cap. (mL)
|
L x diam. (mm)
|
Catalog Number
|
| Complete |
|
20
|
50 x 28
|
|
|
60
|
100 x 32
|
|
|
125
|
105 x 45
|
|
|
250
|
155 x 51
|
|
|
500
|
220 x 70
|
|
| Vessel only |
|
20
|
-
|
|
|
60
|
-
|
|
|
125
|
-
|
|
|
250
|
| |
| |
-
|
|
|
500
|
-
|
|
|
Back to Top
Peptide vessels
 |
Cylindrical style vessels have a 32mm threaded neck on each end that accommodate PTFE valve assembly, Cat. No. Z41,898-6. Caps, adapters, and valves sold separately. (Corning 46810)
|
Approx. cap. (mL)
|
L x diam. (mm)
|
Side port
|
Catalog Number
|
|
20
|
50 x 28
|
14/20
|
|
|
60
|
100 x 32
|
14/20
|
|
|
125
|
105 x 45
|
14/20
|
|
|
250
|
155 x 51
|
14/20
|
|
|
500
|
220 x 70
|
14/20
|
|
|
20
|
50 x 28
|
GL 18*
|
|
|
60
|
100 x 32
|
GL 18*
|
|
|
125
|
105 x 45
|
GL 18*
|
|
|
250
|
155 x 51
|
GL 18*
|
|
|
500
|
205 x 64
|
GL 18*
|
|
|
20
|
50 x 28
|
None
|
|
|
60
|
100 x 32
|
None
|
|
|
125
|
105 x 45
|
None
|
|
|
250
|
155 x 51
|
None
|
|
|
500
|
220 x 70
|
None
|
|
|
*For side port caps, see Cat. No. Z41,697-5 or Z41,701-7 in the System 45 section.
Back to Top
Jacketed vessels
 |
Cylindrical style vessel is jacketed. 32mm screw threads on each end accommodate PTFE valve assembly, Cat. No. Z41,898-6. Supplied with two quick-disconnect fittings for easy assembly and disassembly of hoses. Caps sold separately. (Corning 46812)
|
Cap. (mL)
|
Jacket L (mm)
|
Catalog Number
|
|
20
|
60
|
|
|
60
|
105
|
|
|
125
|
110
|
|
|
250
|
160
|
|
|
500
|
210
|
|
Replacement parts for System 45 peptide vessels |
Adapter, PTFE, for 20mm fritted disc, supplied without disc |
|
| Valve top, PTFE, straight through |
|
| Valve bottom, PTFE |
|
| Fritted disc, coarse, 20mm |
|
| Valve assembly, PTFE, (Includes PTFE adapter, valve bottom, fritted disc, and open-top cap) |
|
|
Back to Top
|
Description
|
Catalog Number
|
| 1/4-28 Male to 1/4-28 male |
|
| 1/4-28 Male to male Luer |
|
Back to Top
Vessels with removable fritted disc
Cylindrical style vessel has a 2mm PTFE valve at the top. The 32mm threaded bottom valve assembly incorporates System 45 technology. The coarse fritted disc is easily removed from the PTFE adapter for cleaning and/or replacement. PTFE top and bottom valves require 1/4- turn to open or close. Valve stems have a 1/4-28 internal thread in the stem which allows a variety of adapters to be used. For male 1/4-28 and male Luer adapters, see Cat. No. Z41,741-6 and Z41,742-4 above. (Corning 46806)
|