|
Glassware Catalog
Peptide Synthesis Flasks
|
Flasks • Funnels • Vessels
|
On this page: Solid-phase peptide synthesis flasks (side joints and a center joint) Solid-phase peptide synthesis flask (Large 85mm top and bottom openings)
Solid-phase peptide synthesis flasks
 |
Has three S.T. 24/40 side joints and a center joint that varies in size according to the capacity of the flask. Coarse frit (25 to 50 mm) and PTFE Rotaflo stopcock permit rapid filtration and washing of resin.
|
Back to Top
Solid-phase peptide synthesis flasks
Large 85mm top and bottom openings allow for easy access when loading and cleaning. Features include a removable fritted disc and a large bore1/4-turn bottom valve for rapid filtration and washing. Two additional ports are provided for reactant addition or gas inlet. Complete assembly includes top cap, funnel, 90mm coarse frit, stirrer assembly, 3ft of 1/4in. PTFE tubing, 1/4in. nut and ferrule. Order stirrer rod and blade separately (below).
|
Cap. (mL)
|
Catalog Number
|
|
500
|
|
|
1,000
|
|
|
2,000
|
|
|
5,000
|
|
Accessories |
|
| Stirrer rod |
|
| PTFE stirrer blade |
|
| Fritted disc, coarse |
|
| Fritted disc, medium |
|
| Fritted disc, fine |
|
| Quick-disconnect hose fitting |