Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Glassware Catalog
Sublimation Equipment - Sigma-Aldrich Glassware Catalog
On this page:
Micro sublimation apparatusSublimation apparatusModified sublimation apparatus
Micro sublimation apparatus
S.T. 24/40 joint, 2mm glass stopcock with O-ring.
Z12,959-3
Recovery flasks for Z12,959-3
100mL
Z51,557-4
Back to top
Sublimation apparatus
Flat-bottom on inner tube for easy sample removal. 2mm glass stopcock.
Starting material capacity
Catalog Number
Up to 3g
Up to 12g
Back to top
Modified sublimation apparatus
Removable vacuum inlet with 2mm glass stopcock improves handling during sample removal and reduces breakage of glassware.
Starting material capacity
Catalog Number
Up to 20g
Back to top