|
Glassware Catalog
System 45
|
Introduction • Step 1 - Select Flasks • Step 2 - Select PTFE Adapters • Step 3 - Select Accessories
|
On this page: Introduction • 3-Step Selection Process • System 45 Compatible Products
 |
Glassware Designed for Organic Chemistry
System 45 is an integrated system of flasks, glassware components, and patented PTFE adapters that incorporate the best in design, safety, and modern materials. System 45 is compatible with existing glassware, and can be phased-in for immediate benefits with minimal replacement expense.
Features:
- Wide GL 32 or GL 45 screw thread flask openings
- Greaseless, gas-tight, 100% freeze-proof seals
- Chemically inert, glass-to-PTFE connections throughout
- Quick-disconnect tubing fittings
|
Back to Top
Simple 3-Step Selection Process
The products in this section of the website are arranged in the general sequence shown below. To locate more System 45 compatible products that appear elsewhere in this website, consult the table at the bottom of this page.
 |
STEP 1 • Select Flasks
Ground-glass joints and glass stoppers have been replaced by wide i.d. screw thread openings and PTFE-lined caps on System 45 flasks for easy access to contents and increased safety. Choose from round-bottom, Kjeldahl, pear-shaped, and straight-sided types.
|
 |
STEP 2 • Select PTFE Adapters
All connections to System 45 flasks are made with patented PTFE adapters. A simple twist of the adapter cap results in a gas-tight, 100% freeze-proof seal without grease. The S.T. joint adapters permit the use of existing lab glassware with System 45 flasks.
|
 |
STEP 3 • Select Components/Accessories
Choose from a wide range of glass System 45 products. See below for a listing of specialty System 45 compatible products located elsewhere in the website.