Hettich Selection Guide
EBA-20 (115 volts)
Hettich Product No. 2002-01
Sigma-Aldrich Product No. Z601039
Select an available tube type below to view volume, tube quantity and centrifuge accessory information
Available Tube types:
Blood / Urine Conical Conical / Skirted Culture Plate Microcentrifuge Microtiter Plate PCR Strips Round Bottom
Blood / Urine
Volume (mL) Tube Quantity Hettich Rotor
Product No.
Sigma-Aldrich Rotor
Product No.
Hettich Adaptor
Product No.
Sigma-Aldrich Adaptor
Product No.
Hettich Bucket
Product No.
Sigma-Aldrich Bucket
Product No.
1.1-1.4 8 Rotor Included Rotor Included 1054-A Z724408 N/A N/A
2.6-2.9 8 Rotor Included Rotor Included 1054-A Z724408 N/A N/A
2.7-3.0 8 Rotor Included Rotor Included 1054-A Z724408 N/A N/A
4.9 8 Rotor Included Rotor Included N/A N/A N/A N/A
4.5-5 8 Rotor Included Rotor Included N/A N/A N/A N/A
5.0 8 Rotor Included Rotor Included 1054-A Z724408 N/A N/A
4.0-7.0 8 Rotor Included Rotor Included N/A N/A N/A N/A
8.0 8 Rotor Included Rotor Included N/A N/A N/A N/A
7.5-8.2 8 Rotor Included Rotor Included N/A N/A N/A N/A
9.0-10.0 8 Rotor Included Rotor Included N/A N/A N/A N/A
8.5-10.0 8 Rotor Included Rotor Included N/A N/A N/A N/A
10.0 8 Rotor Included Rotor Included N/A N/A N/A N/A