Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Hettich Selection Guide
EBA-21 (115 Volts)
Hettich Product No. 1004-01
Sigma-Aldrich Product No. Z601136
Select an available tube type below to view volume, tube quantity and centrifuge accessory information
Available Tube types:
Blood / Urine Conical Conical / Skirted Culture Plate Microcentrifuge Microtiter Plate PCR Strips Round Bottom
Blood / Urine
Volume (mL) Tube Quantity Hettich Rotor
Product No.
Sigma-Aldrich Rotor
Product No.
Hettich Adaptor
Product No.
Sigma-Aldrich Adaptor
Product No.
Hettich Bucket
Product No.
Sigma-Aldrich Bucket
Product No.
1.1-1.4 6 1115 Z601217  1054-A Z724408 1123 Z601225
2.6-2.9 6 1115 Z601217  1054-A Z724408 1123 Z601225
2.7-3.0 6 1115 Z601217  1054-A Z724408 1123 Z601225
4.9 6 1115 Z601217  N/A N/A 1123 Z601225
4.0-5.5 6 1115 Z601217  N/A N/A 1123 Z601225
7.5-8.2 6 1115 Z601217  N/A N/A 1123 Z601225
9.0-10.0 6 1115 Z601217  N/A N/A 1123 Z601225