Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Hettich Selection Guide
Mikro-120 (115 Volts)
Hettich Product No. 1204-01
Sigma-Aldrich Product No. Z720097
Select an available tube type below to view volume, tube quantity and centrifuge accessory information
Available Tube types:
Blood / Urine Conical Conical / Skirted Culture Plate Microcentrifuge Microtiter Plate PCR Strips Round Bottom
Microcentrifuge
Volume (mL) Tube Quantity Hettich Rotor
Product No.
Sigma-Aldrich Rotor
Product No.
Hettich Adaptor
Product No.
Sigma-Aldrich Adaptor
Product No.
Hettich Bucket
Product No.
Sigma-Aldrich Bucket
Product No.
0.2 12 1212 Z727482 2024 Z601209 N/A N/A
0.4 12 1212 Z727482 2024 Z601209 N/A N/A
0.5 12 1212 Z727482 2023 Z601195 N/A N/A
0.8 12 1212 Z727482 2023 Z601195 N/A N/A
1.5 12 1212 Z727482 2031 Z652229 N/A N/A
2.0 12 1212 Z727482 N/A N/A N/A N/A
0.2 18 1218 Z727598 2024 Z601209 N/A N/A
0.4 18 1218 Z727598 2024 Z601209 N/A N/A
0.5 18 1218 Z727598 2023 Z601195 N/A N/A
0.8 18 1218 Z727598 2023 Z601195 N/A N/A
1.5 18 1218 Z727598 2031 Z652229 N/A N/A
2.0 18 1218 Z727598 N/A N/A N/A N/A
0.2 24 1224 Z727709 2024 Z601209 N/A N/A
0.4 24 1224 Z727709 2024 Z601209 N/A N/A
0.5 24 1224 Z727709 2023 Z601195 N/A N/A
0.8 24 1224 Z727709 2023 Z601195 N/A N/A
1.5 24 1224 Z727709 2031 Z652229 N/A N/A
2.0 24 1224 Z652148 N/A N/A N/A N/A