Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Hettich Selection Guide
Mikro-200 (115 Volts)
Hettich Product No. 2400-01
Sigma-Aldrich Product No. Z652105
Select an available tube type below to view volume, tube quantity and centrifuge accessory information
Available Tube types:
Blood / Urine Conical Conical / Skirted Culture Plate Microcentrifuge Microtiter Plate PCR Strips Round Bottom
Microcentrifuge
Volume (mL) Tube Quantity Hettich Rotor
Product No.
Sigma-Aldrich Rotor
Product No.
Hettich Adaptor
Product No.
Sigma-Aldrich Adaptor
Product No.
Hettich Bucket
Product No.
Sigma-Aldrich Bucket
Product No.
0.2 24 2424 Z652148 2024 Z601209 N/A N/A
0.4 24 2424 Z652148 2024 Z601209 N/A N/A
0.5 24 2424 Z652148 2023 Z601195 N/A N/A
0.8 24 2424 Z652148 2023 Z601195 N/A N/A
1.5 24 2424 Z652148 2031 Z652229 N/A N/A
2.0 24 2424 Z652148 2031 Z652229 N/A N/A
0.5 30 2430 Z652156  N/A N/A N/A N/A