Hettich Selection Guide
Mikro-200R
Hettich Product No. 2405-01
Sigma-Aldrich Product No. Z652121
Select an available tube type below to view volume, tube quantity and centrifuge accessory information
Available Tube types:
Blood / Urine Conical Conical / Skirted Culture Plate Microcentrifuge Microtiter Plate PCR Strips Round Bottom
Microcentrifuge
Volume (mL) Tube Quantity Hettich Rotor
Product No.
Sigma-Aldrich Rotor
Product No.
Hettich Adaptor
Product No.
Sigma-Aldrich Adaptor
Product No.
Hettich Bucket
Product No.
Sigma-Aldrich Bucket
Product No.
0.2 24 2424 Z652148 2024 Z601209 N/A N/A
0.4 24 2424 Z652148 2024 Z601209 N/A N/A
0.5 24 2424 Z652148 2024 Z601209 N/A N/A
0.8 24 2424 Z652148 2024 Z601209 N/A N/A
1.5 24 2424 Z652148 2024 Z601209 N/A N/A
2.0 24 2424 Z652148 2024 Z601209 N/A N/A
0.5 30 2430 Z652156  N/A N/A N/A N/A