Hettich Selection Guide
Mikro-220
Hettich Product No. 2200-01
Sigma-Aldrich Product No. Z723851
Select an available tube type below to view volume, tube quantity and centrifuge accessory information
Available Tube types:
Blood / Urine Conical Conical / Skirted Culture Plate Microcentrifuge Microtiter Plate PCR Strips Round Bottom
Blood / Urine
Volume (mL) Tube Quantity Hettich Rotor
Product No.
Sigma-Aldrich Rotor
Product No.
Hettich Adaptor
Product No.
Sigma-Aldrich Adaptor
Product No.
Hettich Bucket
Product No.
Sigma-Aldrich Bucket
Product No.
10.0 6 1016 Z601381 1635 Z601152 N/A N/A
9.0-10.0 6 1016 Z601381 1635 Z601152 N/A N/A
1.1-1.4 12 1015 Z722758 1054-A Z724408 N/A N/A
2.6-2.9 12 1015 Z722758 1054-A Z724408 N/A N/A
2.7-3.0 12 1015 Z722758 1054-A Z724408 N/A N/A
4.9 12 1015 Z722758 N/A N/A N/A N/A
4.5-5 12 1015 Z722758 N/A N/A N/A N/A
1.6-5.0 12 1015 Z722758 1054-A Z724408 N/A N/A
4.0-7.0 12 1015 Z722758 N/A N/A N/A N/A
7.5-8.2 12 1015 Z722758 N/A N/A N/A N/A
9.0-10.0 12 1015 Z722758 N/A N/A N/A N/A
10.0 12 1015 Z722758 N/A N/A N/A N/A