Hettich Selection Guide
Rotina-380 (115 Volts)
Hettich Product No. 1703-01
Sigma-Aldrich Product No. Z720119
Select an available tube type below to view volume, tube quantity and centrifuge accessory information
Blood/ Urine
Volume (mL) Tube Quantity Hettich Rotor
Product No.
Sigma-Aldrich Rotor
Product No.
Hettich Adaptor
Product No.
Sigma-Aldrich Adaptor
Product No.
Hettich Bucket
Product No.
Sigma-Aldrich Bucket
Product No.
7.5-8.2 6 1720 Z631116 1451 Z653403 N/A N/A
9.0-10.0 6 1720 Z631116 1451 Z653403 N/A N/A
8.5-10.0 6 1720 Z631116 1451 Z653403 N/A N/A
2.6-2.9 20 1724 Z657840 1383 Z653381 1430 Z729132
2.7-3.0 20 1724 Z657840 1383 Z653381 1430 Z729132
4.9 20 1724 Z657840 1383 Z653381 1430 Z729132
4.5-5.0 20 1724 Z657840 1383 Z653381 1430 Z729132
1.6-5.0 20 1724 Z657840 1383 Z653381 1430 Z729132
8.5-10 24 1724 Z657840 5231 Z657875 1427 Z657859
4.0-7.0 20 1724 Z657840 1383 Z653381 1430 Z729132
8.5-10.0 24 1726 Z631108 N/A N/A 1369 Z601462
4.9 60 1726 Z631108 N/A N/A 1741 Z653365
4.5-5.0 36 1726 Z631108 N/A N/A 1741 Z653365
7.5-8.2 42 1726 Z631108 N/A N/A 1742 Z601535
9.0-10.0 42 1726 Z631108 N/A N/A 1742 Z601535
10.0 24 1726 Z631108 N/A N/A 1742 Z601535
8.5-10.0 18 1726 Z631108 N/A N/A 1742 Z601535
7.5-8.2 6 1792 Z720178 1451 Z653403 N/A N/A
9.0-10.0 6 1792 Z720178 1451 Z653403 N/A N/A
10.0 6 1792 Z720178 1451 Z653403 N/A N/A
8.5-10.0 6 1792 Z720178 1451 Z653403 N/A N/A
8.5-10 48 1798 Z720208 5248 Z641987 5051 Z724505