Rotofix 32A (115 Volts)
Hettich Product No. 1206-01
Sigma-Aldrich Product No. Z601446
Select an available tube type below to view volume, tube quantity and centrifuge accessory information
Available Tube types:
Blood / Urine Conical Conical / Skirted Culture Plate Microcentrifuge Microtiter Plate PCR Strips Round Bottom
Blood / Urine
Volume (mL) Tube Quantity Hettich Rotor
Product No.
Sigma-Aldrich Rotor
Product No.
Hettich Adaptor
Product No.
Sigma-Aldrich Adaptor
Product No.
Hettich Bucket
Product No.
Sigma-Aldrich Bucket
Product No.
2.6-2.9 20 1324 Z722790 1383 Z653381 1390 Z728926
2.7-3.0 20 1324 Z722790 1383 Z653381 1390 Z728926
4.9 20 1324 Z722790 1383 Z653381 1390 Z728926
4.5-5.0 20 1324 Z722790 1383 Z653381 1390 Z728926
1.1-1.4 40 1624 Z601527 N/A N/A 1741 Z653365
4.9 40 1624 Z601527 N/A N/A 1741 Z653365
2.6-4.9 20 1624 Z601527 1383 Z653381 1381 Z601543
2.7-5.0 20 1624 Z601527 1383 Z653381 1381 Z601543
1.6-5.0 28 1624 Z601527 N/A N/A 1742 Z601535
4.0-5.5 28 1624 Z601527 N/A N/A 1742 Z601535
1.6-7.0 20 1624 Z601527 1383 Z653381 1381 Z601543
4.0-7.0 28 1624 Z601527 N/A N/A 1742 Z601535
8.5-10.0 16 1624 Z601527 N/A N/A 1369 Z601462
7.5-8.2 6 1620A Z657891 1451 Z653403 N/A N/A
9.0-10.0 6 1620A Z657891 1451 Z653403 N/A N/A
10.0 6 1620A Z657891 1451 Z653403 N/A N/A
8.5-10.0 6 1620A Z657891 1451 Z653403 N/A N/A
7.5-8.2 12 1628 Z665533  1621 Z658014 N/A N/A
8.5-10 12 1628 Z665533  1621 Z658014 N/A N/A