Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Labware > Learning Center > Labware Tutorials > Threaded Ground-glass Joint Tutorial
Labware Tutorials
Threaded Ground-glass Joint Tutorial
Image of a threaded ground-glass joint. Image 1—A complete threaded ground-glass
joint.
  Image of an externally threaded female ground-glass standard taper joint and a corresponding male joint with threaded plastic nut. Image 2—It consists of an externally threaded female ground-glass standard taper joint and a corresponding male joint with threaded plastic nut.
Image of a plastic retaining ring. Image 3—A plastic retaining ring loosens the
joint for safe disassembly.
  Image of a threaded plastic nut. Image 4—The threaded plastic nut secures the joint without the use of clamps.
Image of O-ring seals. Image 5—The O-ring seals above the
ground-glass joint, eliminating the need for grease on the joint.