Image 1—A complete threaded ground-glass joint. |
|
Image 2—It consists of an externally threaded female ground-glass standard taper joint and a corresponding male joint with threaded plastic nut. |
| |
Image 3—A plastic retaining ring loosens the joint for safe disassembly. |
|
Image 4—The threaded plastic nut secures the joint without the use of clamps. |
| |
Image 5—The O-ring seals above the ground-glass joint, eliminating the need for grease on the joint. |