Labware > Learning Center > Aldrich Spectral Viewer > Instructional Edition
Labware
Instructional Edition
Aldrich Spectral Viewer Instructional Edition is an electronic reference book on CD-ROM that contains FT-IR and FT-NMR spectra for over 500 representative compounds from a variety of chemical functional groups. Users can practice spectral interpretation by comparing chemical shifts and absorptions for related compounds.
The Instructional Edition Contains:
  • 506 total compounds
  • FT-IR spectra for 428 compounds
  • 300 MHz1H FT-NMR spectra for 437 compounds
  • 75 MHz13C FT-NMR spectra for 440 compounds
  • Both FT-IR and FT-NMR spectra for 359 compounds
Spectral Viewer Program Features:
  • Displays FT-NMR and FT-IR Spectra along with chemical structure and technical data
  • Text Searching by Chemical Name, Aldrich Catalog Number, CAS Number, and Molecular Formula
  • Data Field Searching on Chemical Category and physical properties such as Boiling Point, Melting Point, Density, Flash Point, Molecular Weight, and Refractive Index
  • Spectral Processing using Zoom, Peak Labeling, and Peak Integration (FT-NMR only)
  • Prints Spectra, structure, technical data, peak listings, and integral table (FT-NMR only)
  • Exports Spectrato a file (Bitmap, Metafile, or JPEG formats) or to the clipboard (Bitmap or Metafile formats)
  • Web Link Button provides additional technical data from the Sigma-Aldrich website
spectral_viewer_pic_1
spectral_viewer_pic_2
spectral_viewer_pic_3
Version Cat. No. Qty
Instructional Edition, single user Z53,643-1 1
Instructional Edition, single user Z53,643-1 10 pack
Instructional Edition, Network Z53,633-4 1
For more information email labequipment@sial.com.
System Requirements: Pentium 200MHz, Windows 95 / 98 / NT4-SP5 / 2000 / ME / XP, CD-ROM drive, 300MB hard disk space, 64MB RAM, disk space will vary with version installed.
Spectral Viewer is a trademark and Aldrich is a registered trademark of Sigma-Aldrich Biotechnology, LP
Chemical Abstracts is a registered trademark of the American Chemical Society
Pentium is a registered trademark of Intel Corp.
Windows is a registered trademark of Microsoft Corp.