|
Aldrich Spectral Viewer
FT-NMR Library
|
The Aldrich FT-NMR Library with Spectral Viewer on CD-ROM is an electronic book that contains high-resolution 300MHz 1H and 75MHz 13C spectra for the more than 11,800 compounds found in the printed version of The Aldrich Library of 13C and 1H FT-NMR Spectra. A supplemental library is also available that includes spectra for 3,500 additional compounds not included in the original printed version. Order both the standard and the supplemental library for access to spectra on over 15,000 compounds.
|
 |
 |
Ideal for:
- Teaching students NMR spectral interpretation
- Referencing spectra for comparison studies and the identification of unknown compounds
- Exporting spectra quickly for presentations and papers
|
Features:
- Displays 1H and 13C spectra, integral, chemical structure, and technical data
- Text Searching by Chemical Name, Aldrich Catalog Number, CAS Number, and Molecular Formula
- Advanced data field searching on Chemical Category and physical properties such as Boiling Point, Melting Point, Density, Flash Point, Molecular Weight, and Refractive Index
- Spectra processing using Zoom, Peak Integration, and Peak Labeling
- Print spectra, structure, technical data, integral table and peak listing
- Export spectra to a file (Bitmap, Metafile, or JPEG formats) or to the clipboard (Bitmap or Metafile formats)
- Web link button provides additional technical data from the Sigma-Aldrich Website
Minimum system requirements: Pentium. 200MHz, Windows. 95/98/ME/NT4-SP5/2000, CD-ROM drive, 600MB hard disk space, 64MB RAM
Aldrich is a registered trademark of Sigma-Aldrich Co. Pentium is a registered trademark of Intel Corp. Windows is a registered trademark of Microsoft Corp.
To order, simply click on the catalog number of the version you wish to purchase:
| Version |
FT-NMR Standard library (11,800 compounds) |
FT-NMR Supplemental library (3,500 compounds) |
| Single user, commercial |
Z54,126-5 |
Z53,808-6 |
| Single user, academic |
Z54,138-9 |
Z53,818-3 |
| Network version |
Z54,149-4 |
Z53,797-7 |
| Demonstration program |
Z54,159-1 |
|
| 3-Volume book set |
Z23,103-7 |
|
Clicking on the product number links above will take you the corresponding product detail page, from which you may place your order.