Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Labware > Products > Chemical Manager Software > Component Selection Guide
Labware
Component Selection Guide
< 1 2 3 4 5
The following is a guide to help define which Aldrich® Chemical Manager Software components are needed for your application.
Component Definitions Hardware Configuration - User Driven
Hardware Configuration - Location Driven
Hardware Configuration - Company Driven
Software Configuration - Share Inventory
Software Configuration - Separate Inventory
Component Definitions
Z566934 — Aldrich® Chemical Manager Software, single seat, single computer, 1 year subscription
  • Stand-alone, base software license
  • Single seat defined as software installed on one computer
  • Operates independently or with add-on software licenses (Z567086)
  • Unlimited add-on software licenses (Z567086) can be networked
  • Unlimited users (chemist, technician, assistant, student, instructor, stockroom mngr, etc.)
  • Unlimited storage locations (stockrooms, laboratories, cabinets, etc.)
  • Unlimited inventory (chemicals, lab samples, etc.)
Z567086 — Aldrich® Chemical Manager Software, additional single seat license
  • Add-on software license
  • Single seat defined as software installed on one computer
  • Computer must be networked with computer running base software license (Z566934)
  • Does not function independent of base software license (Z566934)
  • Each additional networked computer requires a separate add-on software license (Z567086)
Z567493 — Aldrich® Chemical Manager Software, custom
  • When integrating data into existing business systems
  • When unique reporting requirements need to be displayed
  • When unique system requirements need to be met
Z566942 — Laser data collector scanner for 1-dimensional barcodes1-dimensional barcodes
  • Hardware used to collect inventory movement
  • Hardware downloads data to software (Z566934 and Z567086)
  • Captures 1-dimensional barcodes (shown to right)
  • It is suggested that each system user have a data collector
Z567620 — Laser data collector scanner for 2-dimensional barcodes2-dimensional barcodes
  • Required for reading Sigma-Aldrich labels
  • Hardware used to collect inventory movement
  • Hardware downloads data to software (Z566934 and Z567086)
  • Captures 1-dimensional barcodes
  • Captures 2-dimensional barcodes (shows to right)
  • It is suggested that each system user have a data collector
Z567639 — Barcode label printer
  • Used to print unique 1-dimensional barcodes onto adhesive barcode labels
  • Labels can be applied to chemical samples
  • Labels can be applied to any product tracked by software (Z566934 or Z567086)
  • Uses Barcode labels (Z567655)
  • Uses Printer ribbon (Z567647)
back to top
Hardware Configuration - User Driven
Aldrich® Chemical Manager Software shown with one data collection device per user. This allows the most flexibility.
Hardware Configuration - User Driven
back to top
Hardware Configuration - Location Driven
Aldrich® Chemical Manager Software shown with one data collection device per location (shared amongst users).
Hardware Configuration - Location Driven
back to top
Hardware Configuration - Company Driven
Aldrich® Chemical Manager Software shown with one data collection device for the entire company (shared amongst users and locations).
Hardware Configuration - Company Driven
back to top
Software Configuration - Share Inventory
Aldrich® Chemical Manager Software shown on four computers that share inventory data.
Software Configuration - Share Inventory
back to top
Software Configuration - Separate Inventory
Aldrich® Chemical Manager Software shown on four computers that have separate inventory data.
Software Configuration - Separate Inventory
back to top
< 1 2 3 4 5