|
< 1 2 3 4 5
The following is a guide to help define which Aldrich® Chemical Manager Software components are needed for your application.
Component Definitions
Z566934 — Aldrich® Chemical Manager Software, single seat, single computer, 1 year subscription
- Stand-alone, base software license
- Single seat defined as software installed on one computer
- Operates independently or with add-on software licenses (Z567086)
- Unlimited add-on software licenses (Z567086) can be networked
- Unlimited users (chemist, technician, assistant, student, instructor, stockroom mngr, etc.)
- Unlimited storage locations (stockrooms, laboratories, cabinets, etc.)
- Unlimited inventory (chemicals, lab samples, etc.)
Z567086 — Aldrich® Chemical Manager Software, additional single seat license
- Add-on software license
- Single seat defined as software installed on one computer
- Computer must be networked with computer running base software license (Z566934)
- Does not function independent of base software license (Z566934)
- Each additional networked computer requires a separate add-on software license (Z567086)
Z567493 — Aldrich® Chemical Manager Software, custom
- When integrating data into existing business systems
- When unique reporting requirements need to be displayed
- When unique system requirements need to be met
Z566942 — Laser data collector scanner for 1-dimensional barcodes
- Hardware used to collect inventory movement
- Hardware downloads data to software (Z566934 and Z567086)
- Captures 1-dimensional barcodes (shown to right)
- It is suggested that each system user have a data collector
Z567620 — Laser data collector scanner for 2-dimensional barcodes
- Required for reading Sigma-Aldrich labels
- Hardware used to collect inventory movement
- Hardware downloads data to software (Z566934 and Z567086)
- Captures 1-dimensional barcodes
- Captures 2-dimensional barcodes (shows to right)
- It is suggested that each system user have a data collector
Z567639 — Barcode label printer
- Used to print unique 1-dimensional barcodes onto adhesive barcode labels
- Labels can be applied to chemical samples
- Labels can be applied to any product tracked by software (Z566934 or Z567086)
- Uses Barcode labels (Z567655)
- Uses Printer ribbon (Z567647)
back to top
Hardware Configuration - User DrivenAldrich® Chemical Manager Software shown with one data collection device per user. This allows the most flexibility.
back to top
Hardware Configuration - Location DrivenAldrich® Chemical Manager Software shown with one data collection device per location (shared amongst users).
back to top
Hardware Configuration - Company DrivenAldrich® Chemical Manager Software shown with one data collection device for the entire company (shared amongst users and locations).
back to top
Software Configuration - Share InventoryAldrich® Chemical Manager Software shown on four computers that share inventory data.
back to top
Software Configuration - Separate InventoryAldrich® Chemical Manager Software shown on four computers that have separate inventory data.
back to top
< 1 2 3 4 5
|