Sigma-Aldrich
Labware > Products > Chemical Manager Software > Overview - Consuming and Reporting
Labware
Overview - Consuming and Reporting
< 1 2 3 4 5 > >>
Consuming Lab Chemicals and Samples
To consume a product that is being managed, use the portable handheld scanner to scan the consume barcode.
Barcode Consume
Then scan the quantity consumed. If you consumed 250mL, scan a 2, 5, and 0.
Barcode - 2
Barcode - 5
Barcode - 0
Next, scan the unit barcode. The software allows you to consume all or portions of product in inventory.
Next, scan the product barcode.
Barcode units
500 mL Barcode
Plug in scanner and download.
The specific product's quantity is removed from your inventory:
Inventory Detail - after
Reporting – Inventory Movement
Additional reports allow the movement of lab chemicals and samples to be logged. Differentiation can be made regarding location and employee.
Inventory movement
Reporting – Inventory Summary by Type
Inventory summary
Reporting – Inventory Detail by Location
Inventory detail by location
< 1 2 3 4 5 > >>