Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Labware > Products > Chemical Manager Software > Overview - Inventory and Barcodes
Labware
Overview - Inventory and Barcodes
< 1 2 3 4 5 > >>
Inducting Existing Inventory
After Aldrich Chemical Manager Software is installed at your facility, existing lab chemicals and sample inventory needs to be manually inducted into the software. This can be done through the easy-to-use entry point show below. Once existing inventory is inducted, all future inventory receiving, consumption, and movement will be accomplished using barcodes.
Inventory receive
Printing Inventory Barcodes
Barcodes are used to identify and differentiate your lab chemicals and samples. Barcodes supplied on containers by manufacturers can be used or barcodes can be created & printed from the software.
Aldrich Inventory
Barcode Label
500 mL Barcode
Prints onto an adhesive barcode label that can be placed directly onto lab chemical container or sample.
Managing Materials using Action Barcodes
There are several action barcodes produced by the software that allow you to manage your inventory:
  • Inbound - Receive and Create
  • Outbound - Consume and Dispose
  • Manage - Count, Audit, and Move
Barcode List Worksheet
Receiving Lab Chemicals and Samples
To receive a product into inventory, use the portable handheld scanner to scan the receive barcode.
 Handheld Scanner    Barcode - Receive
   500 mL Barcode
Plug in scanner and download.
The product is then being managed:
Inventory Detail
< 1 2 3 4 5 > >>