Sigma-Aldrich
Labware > Products > Kodak BioMax Film
Labware
Kodak BioMax Film
Kodak has been the world leader in the development and manufacture of Autoradiography films for more than 30 years. Kodak films are designed specifically for Autoradiography and Chemiluminescence purposes and Kodak scientists have developed their T-Grain technology, being flatter, more tabular silver halide grains used in some of their BioMax product range, that intercept more radiation, providing enhanced sensitivity, speed and less noticeable grain in the film. With development of intensifying screens, including the innovative Transcreen, Kodak further improved the sensitivity, image quality and resolution that laboratory scientists need.
Sigma-Aldrich has been supplying Kodak Autoradiography and Chemiluminescence products for many years, supporting them with high stock levels, sophisticated logistics, easy-to-use order systems, and fast delivery to provide scientists easy access to the best products for their science. See the focus areas below for an easy-to-read product selection guide.
  • Autoradiography & Chemiluminescence Films
  • Electron Micrography Films
  • Autoradiography Accessories
  • Processing Chemicals for Autoradiography Films
Kodak Autoradioragphy & Chemiluminescence Films
BioMax Light
Chemiluminescence Film
A double emulsion (double-sided) film that provides maximum clarity and sensitivity for detection chemiluminescence. Produces highest signal-to-noise ratio of any Kodak film for detecting chemiluminescence and suitable for use with all chemiluminescence kits.
Packaging Size(in.) size(cm.) Kodak No. Sigma-Aldrich Cat. No.
Light-1: Sheets of film with no interleaved paper. 5 x 7 13 x 18 868 9358 Z370398-50EA
7 x 9.5 18 x 24 819 4540 Z373508-50EA
8 x 10 20.3 x 25.4 178 8207 Z370371-50EA
9.5 x 11.8 24 x 30 153 8966 Z373516-50EA
14 x 17 35 x 43 862 5170 Z373494-50EA
Light-2: Each Sheet wrapped in light-tight envelope. 5 x 7 13 x 18 191 7012 Z370428-50EA
8 x 10 20.3 x 25.4 876 1520 Z370401-50EA
Back to top
BioMax MR
Maximum Resolution
A single emulsion (single-sided) film that provides maximum resolution and clearest background. Makes readings and digital scanning autoradiograms from sequencing gels easy and accurate.
The most sensitive film for direct 14C, 35S and 33P autoradiography. Higher resolution than BioMax XAR and MS for 32P and 125I direct and screen exposures, but with reduced sensitivity.
Packaging Size(in.) size(cm.) Kodak No. Sigma-Aldrich Cat. No.
MR-1:  Sheets of film with no interleaved paper 5 x 7 13 x 18 894 1114 Z350397-50EA
7 x 9.5 18 x 24 873 6936 Z353949-50EA
7 x 17 18 x 43 875 6447 Z353957-50EA
8 x 10 20.3 x 25.4 870 1302 Z350370-50EA
9.5 x 11.8 24 x 30 891 2560 Z350427-50EA
11.8 x 15.8 30 x 40 892 9655 Z358478-50EA
14 x 17 35 x 43 871 5187 Z350419-50EA
MR-2: Each sheet wrapped in light-tight envelope 5 x 7 13 x 18 857 2786 Z350427-50EA
8 x 10 20.3 x 25.4 895 2855 Z350400-50EA
14 x 17 35 x 43 856 7232 Z350419-50EA
Back to top
BioMax MS
Maximum Sensitivity
A double emulsion (double-sided) film on clear base. Extremely sensitive to the light emitted from BioMax intensifying screens. When used with intensifying screens, provides maximum sensitivity for 32P, 125I and other isotopes in Northern, Southern and Dot blot applications. Ideal for low intensity samples or when shorter exposure times needed.
Packaging Size(in.) size(cm.) Kodak No. Sigma-Aldrich Cat. No.
MS-1:  Sheets of film with no interleaved paper 5 x 7 13 x 18 894 1114 Z350397-50EA
7 x 9.5 18 x 24 873 6936 Z353949-50EA
7 x 17 18 x 43 875 6447 Z353957-50EA
8 x 10 20.3 x 25.4 870 1302 Z350370-50EA
9.5 x 11.8 24 x 30 891 2560 Z350427-50EA
11.8 x 15.8 30 x 40 892 9655 Z358478-50EA
14 x 17 35 x 43 871 5187 Z350419-50EA
MS-2: Each sheet wrapped in light-tight envelope 5 x 7 13 x 18 857 2786 Z350427-50EA
8 x 10 20.3 x 25.4 895 2855 Z350400-50EA
14 x 17 35 x 43 856 7232 Z350419-50EA
Back to top
BioMax XAR
General Purpose
A double emulsion (double-sided) film on clear base. Very versatile, suitable for use with low and high energy isotopes, or chemiluminescence. Sensitivity with high energy isotopes can be increased by using intensifying screens.
Packaging Size(in.) size(cm.) Kodak No. Sigma-Aldrich Cat. No.
XAR: Sheets of film interleaved with sheets of paper 5 x 7 13 x 18 165 1496 F5388-50EA
8 x 10 20.3 x 25.4 165 1496 F5388-50EA
7 x 9.5 18 x 24 853 2655 Z358487-50EA
9.5 x 11.8 24 x 30 150 1451 Z358495-50EA
11.8 x 15.8 30 x 40 150 6955 Z358584-50EA
14 x 17 35 x 43 165 1512 F5263-50EA
XAR-2: Each sheet wrapped in light-tight envelope 5 x 7 13 x 18 166 0760 F5888-50EA
8 x 10 20.3 x 25.4 165 1579 F5763-50EA
14 x 17 35 x 43 165 1678 F5014-50EA
XAR-roll: 25m roll of film   20 x 2500 153 1680 F5138-1EA
Back to top
X-OMAT LS
Economic, blue tint
A double emulsion (double-sided) film on clear base. Extremely sensitive to the light emitted from BioMax intensifying screens. When used with intensifying screens, provides maximum sensitivity for 32P, 125I and other isotopes in Northern, Southern and Dot blot applications. Ideal for low intensity samples or when shorter exposure times needed.
Packaging Size(in.) size(cm.) Kodak No. Sigma-Aldrich Cat. No.
Sheets of film interleaved with sheets of paper 5 x 7 13 x 18 161 1342 F1274-50EA
8 x 10 20.3 x 25.4 864 6770 F1149-50EA
14 x 17 35 x 43 892 6024 F0899-50EA
Back to top
Electron Imaging and Microscopy
Packaging Size(in.) size(cm.) Kodak No. Sigma-Aldrich Cat. No.
Kodak® electron image film SO-163 3.25. × 4 8.26 x 10.16 801 0100 F8145-250EA
2.5 x 3.54 6.5 × 9 167 9273 F8270-100EA
Kodak® electron microscopy film 4489 3 1/4 in. × 4 in 8.26 x 10.16 166 2238 F8395
2.5 x 3.54 6.5 × 9 161 5228 F5426
2.74 x 2.75 7 x 7 160 8124 F8520
3.54 x 4.72 9 x 12   F8177-100EA
BioMax Exposure Cassette
Lightweight, durable, vinyl covered aluminum cassette with locking clip.
Packaging Size(in.) size(cm.) Kodak No. Sigma-Aldrich Cat. No.
When used at ultracold temperatures, allow to come to room temperature before opening. 8 x 10 20.3 x 25.4 820 9140 C4604-1EA
14 x 17 35 x 43 819 7600 C4854-1EA
BioMax MS Intensifying Screen
Description Size(in.) size(cm.) Kodak No. Sigma-Aldrich Cat. No.
Phospor emits green light, closely matched to the spectral sensitivity of BioMax MS film, to provide highest sensitivity for 32P and 125I autoradiography 7 x 9.5 18 x 24 875 7270 Z363189-1EA
8 x 10 20.3 x 25.4 851 8706 Z363154-1EA
9.5 x 11.8 24 x 30 896 7754 Z363197-1EA
11.8 x 15.8 30 x 40 877 0331 Z363200-1EA
14 x 17 35 x 43 898 1706 Z363162-1EA
BioMax MS Exposure Cassette/Intensifying Screen Set
Description Size(in.) size(cm.) Kodak No. Sigma-Aldrich Cat. No.
Contains BioMax Exposure Cassette and BioMax MS Intensifying Screen described above 8 x 10 20.3 x 25.4 875 0499 C4979-1EA
14 x 17 35 x 43 857 8825 C4854-1EA
BioMax Transcreen
Provides improved resolution and maximum sensitivity. Optimised for use with BioMax MS film.
Transcreen system places screen between blot and film, capturing low intensity radiation that does not reach other intensifying screens
Packaging Size(in.) size(cm.) Kodak No. Sigma-Aldrich Cat. No.
LE: Low energy,for 3H, 14C, 33P, 35S, 45Ca, 59FE 8 x 10 20.3 x 25.4 162 2034 Z374253-1EA
14 x 17 35 x 43 866 0789 Z374261-1EA
HE: High energy, for 32P and 125I 8 x 10 20.3 x 25.4 856 3959 Z374318-1EA
14 x 17 35 x 43 881 1457 Z374326-1EA
BioMax MS Exposure Cassette/Transcreen Set
Contains cassette and Transcreen described above
Packaging Size(in.) size(cm.) Kodak No. Sigma-Aldrich Cat. No.
LE: Low energy (as above) 8 x 10 20.3 x 25.4 103 0667 C5479-1EA
HE: High energy (as above) 8 x 10 20.3 x 25.4 835 6800 C5229-1EA
Intensifying screen cleaner
Kodak No: 106 4930
Sigma-Aldrich Cat. No: Z357097-16FLOZ
Kodak Processing Chemicals for Autoradiography Films
For manual processing
Description Kodak No. Sigma-Aldrich Cat. No.
GBX developer/replenisher
Liquid concentrate to make up indicated volume
190 0943 P7042
GBX fixer/replenisher Liquid concentrate to make up indicated volume 190 1875 P7167
For automated processing systems
Packaging Kodak No. Sigma-Aldrich Cat. No.
RP X-OMAT developer/starter
Packet sufficient to start 8 US gal developer
133 2642 Z354112-1PAK
RP X-OMAT developer/replenisher
Packet sufficient to make up indicated volumes
124 9259
895 6799
Z354090-2x5GA
Z719692-4x4L
RP X-OMAT LO fixer/replenisher
Packet sufficient to make up indicated volumes
805 5102 Z354104-2X5GA
Developer system cleaner
Liquid concentrate, makes 6 US gal
101 3176 Z354120-1GA
Fixer/wash system cleaner
Packet sufficient to make up indicated volumes
139 5110 Z354139-2.5GA