Labware
MolyMod Molecular Model Sets
|
|
|
| |
 |
..:: Molymod. Molecular Model Sets ::..
Top quality Molymod molecular model sets are for use by students, teachers, and researchers. The popular Prentice Hall sets for general and organic chemistry utilize Molymod atoms and bond links and may be upgraded or customized with additional model parts.
Scale is 2.8cm =100pm (open style) and 1.45cm = 100pm (compact, semi-space-filling). Sets include standard color-coded plastic atom centers (with bond holes set at the appropriate angles), bonding links, and a plastic storage box.
|
| |
 |
| P and pi bond set |
|
Build custom model sets or upgrade your current sets with Molymod components. Click on the catalog number links for ordering information. |
| Student Sets |
Z54,453-1 Biochemistry |
|
Z24,315-9 General Chemistry (Prentice Hall) |
| Description |
Holes |
Qty. |
|
Description |
Holes |
Qty. |
| Carbon, black |
4
|
12
|
|
Carbon, black |
4
|
6
|
| Hydrogen, white |
1
|
22
|
|
Halogen, green |
1
|
4
|
| Oxygen, red |
2
|
11
|
|
Hydrogen, white |
1
|
10
|
| Nitrogen, blue |
3
|
6
|
|
Nitrogen, blue |
4
|
2
|
| Nitrogen, blue |
4
|
3
|
|
Octahedral, grey |
6
|
2
|
| Sulfur, yellow |
2
|
1
|
|
Oxygen, red |
2
|
4
|
| Phosphorus, purple |
4
|
1
|
|
Sulfur, yellow |
4
|
2
|
| Medium grey links |
|
40
|
|
Trigonal bipyrimid, brown |
5
|
2
|
| Long flexible links |
|
10
|
|
Medium grey links |
|
20
|
| |
|
|
|
Long grey links |
|
6 |
| |
Z24,316-7 Organic Chemistry (Prentice Hall) |
|
Z54,452-3 Organic/Inorganic Chemistry |
| Description |
Holes |
Qty. |
|
Description |
Holes |
Qty. |
| Hydrogen, white |
1
|
28
|
|
Carbon |
4
|
6
|
| Bromine, orange |
1
|
2
|
|
Hydrogen |
1
|
14
|
| Carbon, black |
4
|
14
|
|
Boron |
3
|
1
|
| Chlorine, green |
1
|
8
|
|
Nitrogen |
3
|
1
|
| Iodine, purple |
1
|
2
|
|
Nitrogen |
4
|
2
|
| Nitrogen, blue |
4
|
4
|
|
Oxygen |
2
|
6
|
| Oxygen, red |
2
|
8
|
|
Oxygen |
4
|
1
|
| 1 mm bonds |
|
40
|
|
Sulfur |
2
|
1
|
| 20 mm bonds |
|
40
|
|
Sulfur |
6
|
1
|
| 35 mm bonds |
|
12
|
|
Phosphorus |
5
|
1
|
| |
|
|
|
Phosphorus
|
3
|
1
|
| |
|
|
|
Halogen, green
|
1
|
6
|
| |
|
|
|
Metal (Na)
|
1
|
2
|
| |
|
|
|
Metal (Ca, Mg)
|
2
|
2
|
| |
|
|
|
Metal (Be)
|
2
|
1
|
| |
|
|
|
Metal (Al)
|
3
|
1
|
| |
|
|
|
Metal (Si, Cu)
|
4
|
1
|
| |
|
|
|
Metal
|
6
|
1
|
|
| Classroom Sets |
Z54,571-6 Basic Organic Chemistry (6 sets) |
|
Z54,572-4 Basic Inorganic Chemistry (6 sets) |
| Description |
Holes |
Qty. |
|
Description |
Holes |
Qty. |
| Carbon, black |
4 |
4 |
|
Carbon, black |
4 |
1 |
| Chlorine, green |
1 |
2 |
|
dsp3, brown |
5 |
1 |
| Hydrogen, white |
1 |
10 |
|
Halogen, green |
1 |
2 |
| Nitrogen, blue |
3 |
1 |
|
Hydrogen, white |
1 |
4 |
| Oxygen, red |
2 |
2 |
|
Metal, grey |
6 |
2 |
| 20 mm bonds |
|
20 |
|
Nitrogen, blue |
3 |
2 |
| 35 mm bonds |
|
4 |
|
Oxygen, red |
2 |
3 |
| |
|
|
|
Phosphorus, purple |
3 |
1 |
| |
|
|
|
Sulfur, yellow |
6 |
1 |
| |
|
|
|
Medium grey links |
|
10 |
| |
|
|
|
Medium purple links |
|
4 |
| |
|
|
|
Long flexible links |
|
6 |
|
| Teacher/Researcher Sets |
Z54,448-5 Biochemistry |
|
Z54,447-7 Complex Ions |
| Description |
Holes |
Qty. |
|
Description |
Holes |
Qty. |
| Carbon |
2 |
2 |
|
Carbon |
2 |
6 |
| Carbon |
3 |
24 |
|
Carbon |
3 |
4 |
| Carbon |
4 |
42 |
|
Carbon |
4 |
18 |
| Hydrogen |
1 |
100 |
|
Halogen |
1 |
6 |
| Hydrogen |
2 |
10 |
|
Hydrogen |
1 |
84 |
| Metal |
4 |
1 |
|
Metal |
4 |
2 |
| Metal |
6 |
1 |
|
Metal |
6 |
7 |
| Nitrogen |
2 |
10 |
|
Nitrogen |
1 |
6 |
| Nitrogen |
3 |
12 |
|
Nitrogen |
4 |
18 |
| Nitrogen |
4 |
12 |
|
Oxygen |
1 |
4 |
| Oxygen |
1 |
10 |
|
Oxygen |
2 |
4 |
| Oxygen, 105° |
2 |
20 |
|
Oxygen |
4 |
4 |
| Oxygen, 180° |
2 |
10 |
|
0.5 mm bonds |
|
92 |
| Phosphorus |
4 |
6 |
|
1 mm bonds |
|
84 |
| Sulfur |
2 |
2 |
|
| Short link tool |
|
1 |
|
| 1 mm bonds |
|
150 |
|
| "V" bonds |
|
2 |
|
| |
|
Z54,446-9 Organic/Inorganic Chemistry |
|
Z54,445-0 Organic Chemistry |
| Description |
Holes |
Qty. |
|
Description |
Holes |
Qty. |
| Carbon |
4 |
12 |
|
Carbon |
2 |
2 |
| Carbon |
5 |
6 |
|
Carbon |
3 |
6 |
| Chlorine |
1 |
8 |
|
Carbon |
4 |
24 |
| Hydrogen |
1 |
12 |
|
Carbon |
5 |
6 |
| Hydrogen |
2 |
2 |
|
Chlorine |
1 |
8 |
| Metal |
1 |
4 |
|
Hydrogen |
1 |
40 |
| Metal |
2 |
3 |
|
Metal |
1 |
2 |
| Metal |
3 |
2 |
|
Metal |
2 |
1 |
| Metal |
4 |
4 |
|
Nitrogen |
4 |
4 |
| Metal |
6 |
1 |
|
Oxygen |
2 |
12 |
| Nitrogen |
3 |
4 |
|
Phosphorus |
4 |
1 |
| Nitrogen |
4 |
6 |
|
Sulphur |
4 |
1 |
| Oxygen |
1 |
5 |
|
Sulphur |
2 |
1 |
| Oxygen |
2 |
13 |
|
1 mm bonds |
|
60 |
| Oxygen |
4 |
4 |
|
20 mm bonds |
|
55 |
| Phosphorus |
3 |
2 |
|
35 mm bonds |
|
25 |
| Phosphorus |
4 |
4 |
|
| Phosphorus |
5 |
1 |
|
| Sulphur |
2 |
8 |
|
| Sulphur |
4 |
4 |
|
| Sulphur |
6 |
1 |
|
| 35 mm bonds |
|
36 |
|
| 20 mm bonds, grey |
|
12 |
|
| 20 mm bonds, purple |
|
38 |
|
| |
Z54,449-3 Organic Stereochemistry |
| Description |
Holes |
Qty. |
| Bromine |
1 |
2 |
| Carbon |
4 |
30 |
| Carbon |
5 |
12 |
| Chlorine |
1 |
8 |
| Hydrogen |
1 |
60 |
| Hydrogen |
2 |
4 |
| Iodine |
1 |
2 |
| Metal |
6 |
3 |
| Nitrogen |
4 |
8 |
| Nitrogen |
5 |
4 |
| Oxygen |
1 |
4 |
| Oxygen |
4 |
12 |
| Phosphorus |
4 |
3 |
| Phosphorus |
5 |
2 |
| Sulphur |
2 |
8 |
| Sulphur |
4 |
4 |
| 1 mm bonds |
|
60 |
| 1 mm bonds, opaque |
|
60 |
| 20 mm bonds |
|
40 |
| 35 mm bonds |
|
12 |
| "V" bonds |
|
20 |
|
Molymod, Atomlink, and Molyorbital are trademarks of Spiring Enterprises Limited.