Labware
Atoms
MolyMod_logo Introduction | Atoms | Bonding Links | Crystal Structure Sets | Molyorbital Sets | Organic Sets
..:: Molymod. Atoms ::..
Build custom model sets or upgrade your current sets with Molymod components.
Element Color Holes   Diam. (mm) Angle Cat. No.
Hydrogen White 1
atom_H
17 - Z54,389-6
Halogen Green 1   17 - Z54,391-8
Metal Gray 1   17 - Z54,392-6
Bromine Orange 1   17 - Z54,393-4
Iodine Purple 1   17 - Z54,394-2
Fluorine Lt. Green 1   17 - Z54,395-0
Halogen Green 1   20 - Z54,396-9
Metal Gray 1   20 - Z54,398-5
Oxygen Red 1   20 - Z54,399-3
Nitrogen Blue 1   20 - Z54,400-0
Halogen Green 1   23 - Z54,401-9
Oxygen Red 1   23 - Z54,402-7
Nitrogen Blue 1   23 - Z54,403-5
Hydrogen White 2   17 180.00 Z54,404-3
Oxygen Red 2
atom_O
23 180.00 Z54,405-1
Oxygen Red 2   23 105.00 Z54,407-8
Sulfur Yellow 2
atom_S
23 105.00 Z54,408-6
Metal Gray 2   23 105.00 Z54,409-4
Nitrogen Blue 2   23 105.00 Z54,410-8
Nitrogen Blue 3   23 107.00 Z54,411-6
Phosphorus Purple 3   23 107.00 Z54,412-4
Metal Gray 3
atom_M
23 107.00 Z54,413-2
Carbon Black 3   23 120.00 Z54,414-0
Nitrogen Blue 3
atom_N
23 120.00 Z54,415-9
Carbon Black 4   23 109.50 Z54,416-7
Nitrogen Blue 4   23 109.50 Z54,417-5
Oxygen Red 4
atom_O4
23 109.50 Z54,418-3
Sulfur Yellow 4   23 109.50 Z54,419-1
Metal Gray 4   23 109.50 Z54,420-5
Silicon Gray 4   23 109.50 Z54,421-3
Halogen Green 4   23 109.50 Z54,422-1
Phosphorus Purple 4   23 109.50 Z54,424-8
Phosphorus Purple 5
atom_P
23 120/90 Z54,425-6
Carbon Black 5   23 120/90 Z54,426-4
Nitrogen Blue 5   23 120/90 Z54,427-2
Metal Gray 5   23 120/90 Z54,428-0
Metal Brown 5   23 120/90 Z54,429-9
Oxygen Red 5   23 120/90 Z54,430-2
Metal Gray 6   23 90.00 Z54,431-0
Halogen Green 6
atom_X
23 90.00 Z54,432-9
Sulfur Yellow 6   23 90.00 Z54,433-7
Hydrogen White 1
atomlink_H
16 AtomlinkTM Z54,459-0