Introduction | Atoms | Bonding Links | Crystal Structure Sets | Molyorbital Sets | Organic Sets ..:: Bonding Links ::..
The bond lengths stated in the table are measured from atom to atom. Bonding links are available in various colors and may be used to represent different bond types. For example, gray links can represent covalent bonds, while purple or white links can represent ionic bonds.
|
Flex bond links are used for double and triple bonds.
|
|
Rigid bond links are used for single bonds.
|
|
Short bond links can be used in place of rigid bonds to make compact models.
|
|
V-bond links are used for making planar 5-atom rings when some of the strain in bonding is absorbed by the "V" which can bend slightly.
|
|
Short bond link removal tool