Labware > Products > MolyMod Molecular Model Sets > Organic Molecule Sets
Labware
Organic Molecule Sets
MolyMod_logo Introduction | Atoms | Bonding Links | Crystal Structure Sets | Molyorbital Sets | Organic Sets
..:: Individual Organic Model Sets ::..
The mushroom-shaped hydrogen AtomlinkTM components and the short (non-visible) bonding links provide compact, semi-space-filling models with an internuclear scale of 1.45cm per Angstrom. Polymer models include long, flexible links for demonstration of the mechanism of polymerization.
polypeptide
PVC
TAG
Polypeptide Polyvinyl chloride Fat (tripalmitin)
amino_acid
glucose
polystyrene
7 Amino acid set Glucose Polystyrene
Polymers Set Contents Cat. No.
PVC - polyvinyl chloride 5 monomer units Z54,460-4
Polypropylene 5 monomer units Z54,461-2
Polystyrene 3 monomer units Z54,462-0
Polyester (terylene) 2 monomer units Z54,463-9
Nylon 66 2 monomer units Z54,464-7
Soaps & Detergents Set Contents Cat. No.
Soap 1 molecule Z54,465-5
Detergent 1 molecule Z54,466-3
Sugars Set Contents Cat. No.
Glucose 2 molecules (alpha, beta) Z54,467-1
Sucrose 1 molecule Z54,469-8
Starch or cellulose 3 glucose units Z54,470-1
Fats & Oils Set Contents Cat. No.
Fat (Tripalmitin) 1 molecule Z54,472-8
Saturated fat - stearic acid C18 H36 O2 1 molecule Z54,473-6
Unsaturated fat - oleic acid C18H34O2 1 molecule Z54,474-4
Cholesterol - C27H46O 1 molecule Z54,475-2
Amino Acids Set Contents Cat. No.
7 Amino acid models collection set 7 models (L-config.) Z54,476-0
8 Model amino acid, peptide upgrade 8 models Z54,477-9
20 Amino acid collection set 20 models Z54,544-9
Polypeptides Set Contents Cat. No.
Polypeptide 5 peptide units Z54,545-7
Protein - beta pleated sheet 15 peptide units Z54,546-5
Alpha - helix 15 peptide units Z54,547-3
DNA Set Contents Cat. No.
DNA (includes stand, 11cm H) 2 base-pairs Z54,548-1
DNA (includes stand, 46cm H) 10 base-pairs Z54,550-3