|
You work hard to achieve accuracy and clarity in your research. Trust only the best to present your results. Partnerships with trusted companies such as Kodak and Polaroid make Sigma-Aldrich the premier source for your autoradiography and photography needs. Sigma's broad offering of cameras, films, processors and accessory items provides a complete source for accurate documentation of your efforts.
|
Polaroid Black and White Print Films
Kodak Films for Autoradiography and Chemiluminescence
Polaroid Color Films
Polaroid 35mm Transparency Films
Kodak Professional Black and White Films
Kodak Filters
Kodak Professional Color Films
Developing Tanks
Intensifying Screens without lead blockers
BioMax TranScreen
Intensifying Screen Cleaner
X-ray Exposure Holders
Exposure Cassettes
Exposure Cassettes/Intensifying Screen Sets
Kodak Exposure Cassettes/Intensifying Screen Sets
Kodak Processing Chemicals for Autoradiography Films
Safelight Lamp, Kodak
Safelight Filters, Kodak
Security Bag for X-Ray Film
Autoradiography Pens
|
Drying Rack, Film
Autoradiography Film Binder
Envelopes, Film Filing
Film Hangers
Thermometers
CU-5 Close-Up Camera System, Polaroid
GelCam Camera System, Polaroid
Macro 5 SLR Camera, Polaroid Instant
Micro-SLR Camera, Polaroid
MP-4+ Camera System, Polaroid
Filter Kits for Electrophoresis Photodocumentation
Lenses
Lighting Assemblies
Model 600 Business Edition Camera, Polaroid
Spectra AF Instant Camera System, Polaroid
Poser Processor, 35 mm, Polaroid
Autoprocessor, Polaroid
Slide Mounter, Polaroid, Illuminated
Slide Mounts, Polaroid
Instant Slide Processing Kit, Polaroid
Instant Slide Printer
Camera Stand
|
Polaroid Black and White Print Films
Product Code |
Type |
* |
ISO Daylight Speed (ASA/DIN) |
Contrast |
Comments |
Packaging |
|
F 4638
|
Type 667, 3 1/4 x 4 1/4
|
P
|
3000/36
|
Med.
|
Professional equivalent of Type 107C
|
2 x 10-exp. packs
|
|
F 5265
|
Type 52, 4x5
|
P
|
400/27
|
Med.
|
Fine grain. Extended range. Broad range of applications.
|
20-exp. tray
|
|
F 4388
|
Type 55, 4x5
|
P/N
|
50/18
|
Med.
|
Fine grain print and high resolution negative.
|
20-exp. tray
|
|
F 4513
|
Type 57, 4x5
|
P
|
3000/36
|
Med.
|
High speed. Medium grain.
|
20-exp. tray
|
|
F 0641
|
Type 331, 3x4
|
P
|
400/27
|
Med.
|
Extended range integral film for CRT image recording.
|
20 x 10-exp. packs
|
|
F 9392
|
Type 553, 4x5
|
P
|
800/30
|
Med.
|
Professional quality. Coaterless
|
10-exp. pack
|
|
F 3390
|
Type 665, 3 1/4 x 4 1/4
|
P/N
|
50/18
|
Med.
|
Fine grain print and high resolution negative.
|
10-exp. pack
|
|
F 6402
|
Polapan PRO 100, 3 1/4 x 4 1/4
|
P
|
100/21
|
Med.
|
Coaterless. High resolution print. Replaces Type 664.
|
2 x 10-exp. packs
|
|
F 2274
|
Polapan 400, 4x5
|
P
|
400/27
|
Med.
|
Coaterless. Fine grain. Equivalent to Type 52.
|
20-exp. tray
|
back to top
Polaroid Color Films
Product Code |
Type |
* |
ISO Daylight Speed (ASA/DIN) |
Comments |
Package |
|
Z36,182-8
|
Type 600, 3 1/4 x 4 1/4
|
P
|
640/29
|
High-definition film with integral battery pack for Model 600 Business Edition Camera. Also fits One Step, Cool Cam, PhotoMagic System 2
|
10-exp. pack, 2 x 10-exp. packs
|
|
F 7772
|
Type 990, 3 5/8x 2 7/8
|
P
|
640/29
|
Spectra Film with integral battery pack for Spectra AF camera
|
2 x 10-exp. packs
|
|
F 1399
|
Polacolor PRO 64T, 3 1/4 x 4 1/4
|
P
|
64/19
|
Tungsten (3200K) color balance. Medium grain. Medium contrast.
|
2 x 10-exp. packs
|
|
F 1649
|
Polacolor PRO 64T, 4x5
|
P
|
64/19
|
Tungsten (3200K) color balance. Medium grain. Medium contrast.
|
20-exp. tray
|
|
F 2024
|
Polacolor PRO 100, 3 1/4 x 4 1/4
|
P
|
100/21
|
Balanced for indirect daylight or electronic flash lighting. Recommended instead of Type 59 for professional use.
|
2 x 10-exp. packs
|
|
F 1391
|
Type 339, 3x4
|
P
|
640/29
|
Integral high speed film for CRT image recording.
|
20 x 10-exp. packs
|
|
F 3515
|
Type 669, 3 1/4 x 4 1/4
|
P
|
80/20
|
Medium contrast with extended range for shadows and highlights
|
2 x 10-exp. packs
|
back to top
Polaroid 35mm Transparency Films
Product Code |
Type |
* |
ISO Daylight Speed (ASA/DIN) |
Comments |
Package |
|
F 7138
|
PolaBlue BN-135
|
8/10
|
High
|
White-on-blue negative film for charts, graphs and text
|
12-exp. roll
|
|
F 4763
|
PolaChrome CS-135
|
40/17
|
Med.
|
General purpose color slide material with medium grain
|
12-exp. roll
|
|
F 4013
|
PolaChrome CS-135
|
40/17
|
Med.
|
General purpose color slide material with medium grain
|
36-exp. roll
|
|
F 3888
|
High Contrast PolaChrome HCP-135
|
40/17
|
High
|
For color reproduction of graphics, line art and text
|
12-exp. roll
|
|
F 4888
|
PolaGraph HC-135
|
400/27
|
High
|
B&W positive film for line art and text.
|
12-exp. roll
|
|
F 4138
|
PolaPan CT-135
|
125/22
|
Med.
|
General purpose continuous tone for B&W positive slides.
|
12-exp. roll
|
back to top
Kodak Professional Black and White Films
Product Code |
(Professional) Film Designation |
Contrast |
Process |
Format |
Package |
|
F 1896
|
Ektagraphic
|
High (orthochromatic)
|
D-11
|
Reverse text transparency
|
3 x 36 exp.
|
|
F 2541
|
Technical Pan 2415
|
Variable
|
D-19 or HC-110
|
Negative
|
3 x 36 exp.
|
|
F 2646
|
T-MAX 100
|
Continuous tone
|
D-76, HC-110 or T-MAX
|
Negative
|
3 x 24 exp.
|
|
F 9020
|
T-MAX 100
|
Continuous tone
|
D-76, HC-110 or T-MAX
|
Negative
|
3 x 36 exp.
|
|
F 2771
|
T-MAX 400
|
Continuous tone
|
D-76, HC-110 or T-MAX
|
Negative
|
3 x 24 exp.
|
|
F 2521
|
T-MAX 400
|
Continuous tone
|
D-76, HC-110 or T-MAX
|
Negative
|
3 x 36 exp.
|
back to top
Kodak Filters 75 mm (3 in.) square gelatin filters. Primarily for black-and-white photography intended to vary contrast and tonal reproduction.
Product Code |
Compatibility |
Description |
|
F 9265
|
Wratten 2A
|
Light yellow UV filter for use with the Wratten 22 filter for the photography of ethidium bromide stained nucleic acids.
|
|
F 0146
|
Wratten 11
|
Yellowish green, corrects panchromatic film to tungsten eye response.
|
|
F 0271
|
Wratten 12
|
Deep yellow, minus-blue filter.
|
|
F 8390
|
Wratten 15
|
Deep yellow filter provides optimal contrast for black-and-white photography of Coomassie Brilliant Blue G/R stained proteins.
|
|
F 7272
|
Wratten 22
|
Deep orange filter provides optimal contrast for black-and-white photography of ethidium bromide stained nucleic acids.
|
|
F 0396
|
Wratten 25
|
Red tricolor. Used for color separation and infrared photography.
|
|
F 0521
|
Wratten 27
|
Blue tricolor. Used for color separation.
|
|
F 0646
|
Wratten 58
|
Green tricolor. Used for color separation.
|
A UV filter such as Wratten 2A, used between the light source and the Wratten 22, reduces background and protects the Wratten 22 against photo bleaching.
back to top
Kodak Professional Color Films
Product Code |
(Professional) Film Designation |
Color Balance |
Process |
Format |
Package |
|
F 1021
|
Ektachrome 64T
|
Tungsten
|
E-6
|
Color transparency
|
1 x 36 exp., 3 x 36 exp.
|
|
F 1271
|
Ektachrome 100
|
Daylight
|
E-6
|
Color transparency
|
3 x 36 exp.
|
|
F 1396
|
Ektachrome 100 Plus
|
Daylight
|
E-6
|
Color transparency
|
3 x 36 exp.
|
|
F 1521
|
Ektachrome 160
|
Tungsten
|
E-6
|
Color transparency
|
1 x 36 exp., 3 x 36 exp.
|
|
F 1646
|
Ektachrome 200
|
Daylight
|
E-6
|
Color transparency
|
3 x 36 exp.
|
|
F 1771
|
Ektachrome 400
|
Daylight
|
E-6
|
Color transparency
|
3 x 24 exp.
|
|
F 2021
|
Kodachrome 25
|
Daylight
|
K-14
|
Color transparency
|
3 x 36 exp.
|
|
F 2271
|
Kodachrome 64
|
Daylight
|
K-14
|
Color transparency
|
3 x 36 exp.
|
|
F 2396
|
Kodachrome 200
|
Daylight
|
K-14
|
Color transparency
|
3 x 36 exp.
|
|
F 2896
|
Vericolor III (160)
|
Daylight
|
C-41
|
Color negative
|
5 x 36-exp.
|
back to top
Developing Tanks
Product Code |
Material |
Dimensions |
Features |
|
D 0792
|
Acrylic
|
23h x 13w x 15d in.
|
Three compartments. Drains with hose connectors. Side compartments with acrylic covers. Overflow for constant water circulation. Built-in carrying handles make the unit semi-portable.
|
|
T 1294
|
Stainless Steel
|
20h x 16w x 36d in.
|
All heli-arc welded Type 316 stainless steel. Includes two 5-gal inserts, two insert covers, standpipe drain system, and agitating water inlet. Backed enamel painted steel stand included.
|
back to top
Intensifying Screens without lead blockers
Product Code |
Application |
Packaging |
|
Kodak X-Omatic Regular Phosphar coating on flexible polyester support emits near UV and blue light, optimized for use with Kodak XAR film. Without foam backing.
|
|
Z35,699-9
|
For 8 x 10 in. film
|
2/pkg.
|
|
Z35,701-4
|
For 9.5 x 11.8 in. film
|
2/pkg.
|
|
Z35,700-6
|
For 14 x 17 in. film
|
2/pkg.
|
|
Lite Plus, Sigma An economical alternative to the Dupont Cronex Lightning Plus
|
|
I 5893
|
For 5 x 7 in. and 13 x 18 cm film
|
2/pkg.
|
|
Z36,016-3
|
For 7 x 9.5 in. and 18 x 24 cm film
|
2/pkg.
|
|
Z36,019-8
|
For 7 x 17 in. and 18 x 43 cm film
|
2/pkg.
|
|
I 6018
|
For 8 x 10 in. film
|
2/pkg.
|
|
I 6143
|
For 14 x 17 in. and 35 x 43 cm film
|
2/pkg.
|
|
Z36,017-1
|
For 10 x 12 in. and 24 x 30 cm film
|
2/pkg.
|
|
Z36,020-1
|
For 30 x 40 cm film
|
2/pkg.
|
|
Kodak BioMax MS Phosphar emits green light, closely matched to the spectral sensitivity of BioMax MS film. Provides highest sensitivity for 32P and 125I autoradiography
|
|
Z36,317-0
|
For 5 x 7 in. and 13 x 18 cm film
|
1/pkg.
|
|
Z36,318-9
|
For 7 x 10 in. and 18 x 24 cm film
|
1/pkg.
|
|
Z36,315-4
|
For 8 x 10 in. film
|
1/pkg.
|
|
Z36,319-7
|
For 10 x 12 in. and 24 x 30 cm film
|
1/pkg.
|
|
Z36,316-2
|
For 14 x 17 in. and 35 x 43 cm film
|
1/pkg.
|
|
Z36,320-0
|
For 12 x 16 in. and 30 x 40 cm film
|
1/pkg.
|
|
Rarex Blue III, Sigma A blue emitting, rare-earth screen which is compatible with full or half speed films. The construction of these screens is specially designed to maximize useful life and minimize damage due to moisture and other common contaminants.
|
|
Z37,386-9
|
For 5 x 7 in. and 13 x 18 cm film
|
2/pkg.
|
|
Z37,387-7
|
For 8 x 10 in. film
|
2/pkg.
|
|
Z37,388-5
|
For 14 x 17 in. and 35 x 43 cm film
|
2/pkg.
|
|
Dupont Cronex Quanta III An ultra speed rare-earth intensifying screen for general autoradiographic use. For autoradiography exposures greater than 24 hours, we recommend the Sigma Lite Plus screens listed above.
|
|
I 4636
|
For 5 x 7 in. and 13 x 18 cm film
|
2/pkg.
|
|
I 4886
|
For 14 x 17 in. and 35 x 43 cm film
|
2/pkg.
|
back to top
BioMax TranScreen For enhanced exposures with low energy beta-emitters. Optimized for use with BioMax MS film, but can be used with other films.
Product Code |
High/Low Energy Application |
Size Application |
|
Z37,425-3
|
Low-energy
|
For 8 x 10 in. film, can be cut to fit 18 x 24 cm film
|
|
Z37,428-8
|
Low-energy
|
8 x 10 with BioMax vinyl-clad aluminum cassette
|
|
Z37,426-1
|
Low-energy
|
For 14 x 17 in. and 35 x 43 cm film
|
|
Z37,429-6
|
Low-energy
|
14 x 17 (35 x 43 cm) with BioMax vinyl-clad aluminum cassette
|
|
Z37,431-8
|
High-energy
|
For 8 x 10 in. film, can be cut to fit 18 x 24 cm film
|
|
Z37,433-4
|
High-energy
|
8 x 10 with BioMax vinyl-clad aluminum cassette
|
|
Z37,432-6
|
High-energy
|
For 14 x 17 in. and 35 x 43 cm film
|
|
Z37,434-2
|
High-energy
|
14 x 17 (35 x 43 cm) with BioMax vinyl-clad aluminum cassette
|
back to top
Intensifying Screen Cleaner Removes dust and dirt, and dissipates static charge from intensifying screens.
Product Code |
Description |
Size |
|
I 7888
|
Alcohol-free in dispenser-top bottle for Dupont Cronex III intensifying screens
|
16 fl. oz
|
|
Z35,705-7
|
Recommended for Kodak X-Omatic intensifying screens
|
16 fl. oz
|
back to top
X-ray Exposure Holders Kodak, Vinyl-covered, with Velcro closure Flexible pebbled vinyl outer cover and a Velcro closure for ease of handling in the darkroom. Economical alternative to stainless steel cassettes for exposures down to -70 °C
Product Code |
Application |
|
E 8510
|
For 5 x 7 in. and 13 x 18 cm film
|
|
E 8635
|
For 8 x 10 in. film
|
|
E 8760
|
For 14 x 17 in. and 35 x 43 cm film
|
back to top
Exposure Cassettes Stainless steel, with metal cross-bar closure. For exposing sheet X-ray film during autoradiography. Riveted floating hinges and pressure from cross-bars maintain close contact between film and sample.
Product Code |
Application |
|
E 9260
|
For 5 x 7 in. and 13 x 18 cm film
|
|
Z36,008-2
|
For 7 x 9.5 in. and 18 x 24 cm film
|
|
Z36,010-4
|
For 7 x 17 in. and 18 x 43 cm film
|
|
E 9385
|
For 8 x 10 in. film
|
|
E 9510
|
For 14 x 17 in. and 35 x 43 cm film
|
|
E 9635
|
For 14 x 36 in. film
|
|
Z36,009-0
|
For 24 x 30 cm film
|
|
Z36,011-2
|
For 30 x 40 cm film
|
|
Kodak BioMax Lightweight, vinyl-covered aluminum cassettes. Light-tight with a single locking clip. Bring to room temperature before opening.
|
|
Z36,323-5
|
For 8 x 10 in. and 20 x 25 cm film
|
|
Z36,324-3
|
For 14 x 17 in. and 35 x 43 cm film
|
back to top
Exposure Cassettes/Intensifying Screen Sets The economical alternative. Each set contains one stainless steel exposure cassette and one Sigma Lite Plus intensifying screen.
Product Code |
Application |
|
E 6767
|
For 5 x 7 in. and 13 x 18 cm film
|
|
Z36,012-0
|
For 7 x 9.5 in. and 18 x 24 cm film
|
|
Z36,014-7
|
For 7 x 17 in. and 18 x 43 cm film
|
|
E 6892
|
For 8 x 10 in. film
|
|
E 7017
|
For 14 x 17 in. and 35 x 43 cm film
|
|
Z36,013-9
|
For 24 x 30 cm film
|
|
Z36,015-5
|
For 30 x 40 cm film
|
back to top
Kodak Exposure Cassettes/Intensifying Screen Sets Set contains one Kodak C-2 lightweight and durable vinyl-covered aluminum cassette and two Kodak X-Omatic intensifying screens. Note: When used at low temperature (-70 °C), warm to ambient temperature for 15 minutes before opening to prevent cracking polyurethane hinge.
Product Code |
Application |
|
C 5729
|
For 8 x 10 in. film
|
|
C 5854
|
For 9.5 x 11.8 in. film
|
|
C 5604
|
For 14 x 17 in. film
|
back to top
Kodak Films for Autoradiography and Chemiluminescence
Product Code |
Description |
Size |
Kodak # |
Package |
|
X-OMAT AR
|
|
F 5513
|
XAR-5: Sheets of film interleaved with sheets of paper
|
8 x 10 in.
|
165 1454
|
50 sheets
|
|
F 5763
|
XAR-2 Ready Pack. With high-energy emitters, film may be exposed directly through envelope without cassettes.
|
8 x 10 in.
|
165 1579
|
50 sheets
|
|
F 5888
|
XAR-2 Ready Pack. With high-energy emitters, film may be exposed directly through envelope without cassettes.
|
13 x 18 cm
|
103 6037
|
25 sheets
|
|
F 5014
|
XAR-2 Ready Pack. With high-energy emitters, film may be exposed directly through envelope without cassettes.
|
35 x 43 cm
|
165 1678
|
50 sheets
|
|
F 5388
|
XAR-5: Sheets of film interleaved with sheets of paper
|
13 x 18 cm
|
844 7195
|
25 sheets
|
|
Z35,848-7
|
XAR-5: Sheets of film interleaved with sheets of paper
|
18 x 24 cm
|
853 2665
|
50 sheets
|
|
Z35,849-5
|
XAR-5: Sheets of film interleaved with sheets of paper
|
24 x 30 cm
|
150 1451
|
50 sheets
|
|
Z35,858-4
|
XAR-5: Sheets of film interleaved with sheets of paper
|
30 x 40 cm
|
150 6955
|
50 sheets
|
|
F 5263
|
XAR-5: Sheets of film interleaved with sheets of paper
|
35 x 43 cm
|
165 1512
|
50 sheets
|
|
F 5138
|
XAR-351: Roll film; cut in the darkroom to fit application
|
20 cm x 25 m
|
153 1680
|
1 roll
|
|
BioMax MR Single emulsion film that provides maximum resolution and clearest background. The most sensitive film for direct 14C, 35S, and 33P autoradiography
|
|
Z35,039-7
|
MR-1: Sheets of film with no interleaved paper
|
5 x 7 in.
|
894 1114
|
50 sheets
|
|
Z35,394-9
|
MR-1: Sheets of film with no interleaved paper
|
7 x 9.5 in.
|
873 6936
|
50 sheets
|
|
Z35,395-7
|
MR-1: Sheets of film with no interleaved paper
|
7 x 17 in.
|
875 6447
|
50 sheets
|
|
Z35,037-0
|
MR-1: Sheets of film with no interleaved paper
|
8 x 10 in.
|
870 1302
|
50 sheets
|
|
Z35,846-0
|
MR-1: Sheets of film with no interleaved paper
|
24 x 30 cm
|
891 2560
|
50 sheets
|
|
Z35,847-9
|
MR-1: Sheets of film with no interleaved paper
|
30 x 40 cm
|
892 9655
|
50 sheets
|
|
Z35,038-9
|
MR-1: Sheets of film with no interleaved paper
|
35 x 43 cm
|
871 5187
|
50 sheets
|
|
Z35,042-7
|
MR-2: Each sheet of film is sealed in a light-tight envelope
|
5 x 7 in.
|
820 2160
|
25 sheets
|
|
Z35,040-0
|
MR-2: Each sheet of film is sealed in a light-tight envelope
|
8 x 10 in.
|
895 2855
|
50 sheets
|
|
Z35,041-9
|
MR-2: Each sheet of film is sealed in a light-tight envelope
|
14 x 17 in.
|
856 7232
|
50 sheets
|
|
X-OMAT RA Duplicating Single emulsion film on clear base. For 1:1 reproduction of autoradiograms or stained electrophoresis gels.
|
|
Z35,881-9
|
Sheets of film with no interleaved paper
|
8 x 10 in.
|
140 9760
|
100 sheets
|
|
Z35,882-7
|
Sheets of film with no interleaved paper
|
35 x 43 cm
|
138 9923
|
100 sheets
|
|
X-OMAT LS Coated with emulsion on both sides of a blue-tinted base. Produces higher quality images than XAR film. However, the exposure time is approximately doubled.
|
|
F 1149
|
Kodak life science imaging film equivalent to XRP. Sheets of film interleaved with sheets of paper
|
8 x 10 in.
|
160 4545
|
50 sheets
|
|
F 1274
|
Kodak life science imaging film equivalent to XRP. Sheets of film interleaved with sheets of paper
|
13 x 18 cm
|
103 6227
|
25 sheets
|
|
F 0899
|
Kodak life science imaging film equivalent to XRP. Sheets of film interleaved with sheets of paper
|
35 x 43 cm
|
181 1769
|
50 sheets
|
|
Direct Exposure Film Coated with emulsion on both sides of clear base, with gelatin coating for abrasion resistance.
|
|
F 2774
|
DEF-5: Sheets of film interleaved with sheets of paper
|
13 x 18 cm
|
154 2505
|
50 sheets
|
back to top
Kodak Processing Chemicals for Autoradiography Films
Product Code |
Description |
Reconstituted Volume |
|
For Automated Processing Systems
|
|
Z35,409-0
|
RP X-OMAT Developer/Replenisher (powder)
|
4 x 1 gal., 2 x 5 gal.
|
|
Z35,410-4
|
RP X-OMAT Fixer/Replenisher (powder)
|
4 x 1 gal., 2 x 5 gal.
|
|
Z35,411-2
|
RP X-OMAT Developer Starter (powder)
|
8 gal.
|
|
Z35,412-0
|
Developer System Cleaner (powder)
|
6 gal.
|
|
Z35,413-9
|
Fixer/Wash System Cleaner (powder)
|
2.5 gal.
|
|
For Manual Processing
|
|
P 7042
|
GBX Developer/Replenisher (liquid concentrate)
|
1 gal., 5 gal.
|
|
P 7167
|
GBX Fixer/Replenisher (liquid concentrate)
|
1 gal., 5 gal.
|
|
Z35,414-7
|
GBX Developer and Fixer Twin Pack (liquid concentrate)
|
2 packs that make 1 gal. each.
|
back to top
Safelight Lamp, Kodak
Product Code |
Description |
|
S 2270
|
Model B Adjustable
|
back to top
Safelight Filters, Kodak
Product Code |
Description |
Application |
|
F 9645
|
Type 1, Red
|
For some blue-sensitive materials and most typesetting materials.
|
|
F 9770
|
Type 1A, Light Red
|
For slow orthochromatic materials.
|
|
F 9515
|
Type GBX-2
|
For use with all Kodak autoradiography films with a standard 15W or smaller frosted bulb; with X-OMAT AR, use a 7.5W bulb.
|
|
F 9395
|
Type OA, Greenish Yellow
|
For black-and-white contact and duplicating materials and projection films
|
|
F 9520
|
Type OC, Light Amber
|
For contact and enlarging papers
|
back to top
Security Bag for X-Ray Film Tightly woven, black nylon-coated inside with opaque black vinyl. An overlapping flap closure secures the bag on all edges with a hook-and-loop seal. Protects one box of film.
Product Code |
Application |
Qty |
|
Z38,014-8
|
For 5 x 7 in. film (13 x 18 cm)
|
|
Z38,015-6
|
For 8 x 10 in. film (20 x 25 cm)
|
|
Z38,016-4
|
For 14 x 17 in. film (35 x 43 cm)
|
|
Security Bag Accessories
|
|
L 5149
|
Phosphorescent Label sheets for autoradiograms
|
Ten 4 x 6 inch sheets, may be cut to size
|
|
R 8008
|
Rulers, Fluorescent Two different intensities to match different fluorescent samples.
|
Two 7 x 200 mm and two 7 x 100 mm rulers
|
|
R 8133
|
Rulers, Autoradiography Non-radioactive phosphorescent emission intensity is adjusted to provide sharp markings. Adhesive backing.
|
Four 11 x 200 mm rulers
|
|
I 5263
|
Identi-Kit Non-radioactive phosphorescent, adhesive backed letters, numbers and arrows for use with autoradiography films
|
back to top
Autoradiography Pens Non-radioactive markers for annotating autoradiograms. Bright green ink is visible in normal light, and phosphoresces with an emission peak at 560 nm. Total energy can be adjusted by pre-exposure to room light.
Product Code |
Energy Level |
Application |
|
Z36,351-0
|
Normal
|
For use with Kodak X-OMAT AR or similarly sensitive films
|
|
Z36,352-9
|
High
|
For use with less sensitive films and under conditions that make film less responsive
|
back to top
Drying Rack, Film
Product Code |
Application |
|
D 3411
|
Stainless steel, for use with film hangers. Holds 12 hangers
|
back to top
Autoradiography Film Binder Vinyl-covered, 10-ring loose leaf binder for storing and protecting autoradiography films. Supplied with 10 pre-punched protectors, each of which can accommodate two 14 x 17 inch films. Total capacity is approx. 40 films.
Product Code |
Application |
|
Z37,751-1
|
Binder, includes 10 sheet protectors
|
|
Z37,753-8
|
Additional sheet protectors, package of 10
|
back to top
Envelopes, Film Filing Heavy-duty Kraft envelopes with printed spaces for identification
Product Code |
Application |
Quantity |
|
E 2637
|
For 5 x 7 in. and 13 x 18 cm film
|
100/pkg
|
|
E 2762
|
For 8 x 10 in. film
|
500/pkg
|
|
E 2887
|
For 14 x 17 in. and 35 x 43 cm film
|
100/pkg
|
back to top
Film Hangers For tank development of X-ray film. Stainless steel frame has attached clips to hold the film taut. The holder for 36 in long film holds the film tightly in a U shape to fit a standard tank.
Product Code |
Application |
|
F 6013
|
For 5x7 in. and 13x18 cm film
|
|
F 6138
|
For 8x10 in. film
|
|
F 6263
|
For 14x17 in. and 35x43 cm film
|
|
F 6388
|
For 14x36 in. film
|
|
Storage Rack, Film Hanger
|
|
S 7645
|
Stainless steel, for storage of X-ray film hangers. Holds 12-15 hangers. Screws included.
|
back to top
Thermometers For monitoring temperature in developing tank without thermostatic mixing valve.
Product Code |
Material/Type |
Calibration temperature |
|
T 6655
|
Dial-type, fluorescent
|
50-150 °F (10-65 °C)
|
|
T 6780
|
Glass, floating
|
25-100 °F
|
back to top
CU-5 Close-Up Camera System Accessories, Polaroid
Product Code |
Description |
|
CU-5 Close-Up Camera Accessories
|
|
C 0665
|
Camera body with contour grip
|
|
F 4140
|
Frame attachment for 1:1 pictures
|
|
L 4519
|
Lens/shutter, 3 in, f/4.5, 75mm
|
|
P 4920
|
Power pack, portable AC/DC, 110-120V
|
|
P 7546
|
Power pack, portable AC/DC, 220-240V
|
back to top
GelCam Camera System, Polaroid Used for instant photographic records from light-emitting sources. Hand-held operation. No focusing required.
Product Code |
Description |
|
C 0540
|
GelCam Camera
|
|
GelCam Camera Accessories Oscilloscope Hoods (flat face) with Filter Holder focal plane is flush with the end of the hood for flat-faced oscilloscope screens
|
|
H 8518
|
5 in. diagonal
|
|
H 8643
|
6 in. diagonal
|
|
H 8763
|
7 in. diagonal
|
|
H 8893
|
12 in. diagonal
|
|
Electrophoresis Hoods (flat face) with Filter Holder Focal plane is recessed slightly inside the hood to compensate for typical gel thickness when hood rests on transilluminator
|
|
Z35,968-8
|
5 in. diagonal
|
|
Z35,969-6
|
6 in. diagonal
|
|
Z35,971-8
|
7 in. diagonal
|
|
Z35,972-6
|
13 in. diagonal
|
back to top
Macro 5 SLR Camera, Polaroid Instant
Product Code |
Description |
|
Z37,762-7
|
Close-up system that is easy to use, versatile and portable. Offers 5 magnifications.
|
back to top
Micro-SLR Camera, Polaroid Compatible with virtually all microscopes, no special camera port is required.
Product Code |
Description |
|
Z35,804-5
|
Micro-SLR Camera with 110V AC adaptor
|
|
Z35,805-3
|
Micro-SLR Camera with 220V AC adaptor
|
back to top
MP-4+ Camera System, Polaroid Extremely versatile. Useful for gel documentation, copying, small object or specimen photography, and off-stand photography.
Product Code |
Description |
|
MP-4P
|
MP-4+ Camera, Polaroid
|
|
Parts and Accessories for Polaroid MP-4+
|
|
B 3407
|
Baseboard
|
|
C 3540
|
Camera Head, Sliding
|
|
C 1790
|
Camera Mount, Universal
|
|
MP-CSP
|
Standard Fixed Column
|
|
F 8640
|
Film Holder (3 1/4 x 4 1/4 in. pack film)
|
|
D 8413
|
Dark Slide Replacement
|
|
F 8765
|
Film Holder (4 x 5 in. sheet film)
|
|
G 7395
|
Ground Glass
|
back to top
Filter Kits for Electrophoresis Photodocumentation
Product Code |
Application |
|
MP-4F
|
Color correction for tungsten lamps when using Polaroid color films
|
|
Z60,474-7
|
Contains three filters for filming #15 deep yellow for ethidium bromide, #8 med. yellow for Coomassie Blue, and #58 green for silver.
|
back to top
Lenses
back to top
Lighting Assemblies
Product Code |
Type |
|
MPP-120
|
Standard lighting assembly
|
|
L 6151
|
Standard light arms
|
|
S 1279
|
Standard shields
|
|
B 3532
|
Tungsten floodlight bulbs
|
|
Other Accessories
|
|
M 7777
|
Macro-Extension
|
|
R 6007
|
Reflex viewer and hood
|
|
S 1770
|
Shutter, self-locking Copal
|
back to top
Model 600 Business Edition Camera, Polaroid
Product Code |
Type |
|
Z36,180-1
|
Model 600 Business Edition Camera, Polaroid
|
back to top
Spectra AF Instant Camera System, Polaroid Fully automatic instant camera for aim and shoot ease of use in the lab or field.
Product Code |
Description |
|
C 4929
|
Spectra AF Camera, Polaroid
|
|
C 6178
|
Spectra AF Camera Close-Up Kit
|
|
Spectra AF Camera Accessories
|
|
C 8429
|
Copy stand
|
|
L 4400
|
Close-up lens
|
back to top
Power Processor, 35 mm, Polaroid
Product Code |
Description |
|
P 5045
|
Automatic motorized processor for 35 mm instant slide films
|
back to top
Autoprocessor, Polaroid
Product Code |
Description |
|
A 0546
|
Manually operated processor for 35 mm instant slide films
|
back to top
Slide Mounter, Polaroid, Illuminated
Product Code |
Description |
|
S 0891
|
For 35 mm transparencies
|
back to top
Slide Mounts, Polaroid
Product Code |
Description |
|
S 1016
|
35 mm for manual use or use with slide mounter
|
back to top
Instant Slide Processing Kit, Polaroid
Product Code |
Description |
|
Z36,160-7
|
Power processor, slide mounter, slide mounts
|
back to top
Instant Slide Printer
Product Code |
Description |
|
I 2137
|
Daylab Jr., for 3 1/4 x 4 1/4 prints from any print, transparency or slide
|
back to top
Camera Stand
Product Code |
Description |
|
Z37,763-5
|
Attaches to Polaroid SD-34, CR-10, DS-31 and DS-39 cameras.
|
back to top
|
|
| |
| |
|