|
Reagents and Equipment Procedure References
Reagents and Equipment
- 20x Saline-sodium citrate buffer (SSC: 3 M NaCl, 0.3 M sodium citrate, pH 7, or Product No. S6639).
- RNase A (Product No. R4642) 100 µg/ml in 2x SSC.
- Pepsin (Product No. P6887) 40 units/ml in 10 mM HCl.
- Paraformaldehyde, EM grade (Product No. P6148) freshly depolymerized, 4% w/v in water.
- Ethanol.
- Labeled probe. Plasmid DNA is labeled with biotin-11-dUTP using nick translation random priming or the polymerase chain reaction (e.g., ADVANCE™ Nick Translation Kit).
- Hybridization mix solution: 50% formamide (Product No. F7508), 10% dextran sulfate (Product No. D8906), 0.1% SDS (Product No. L4390), 0.5-1.5 ng/µl labeled probe and 300 ng/ml Salmon Sperm DNA (Product No. D7656) in 2x SSC.
- Wash buffer: 20% formamide (Product No. F7508) in 0.1x SSC.
- Detection buffer: 0.2% Tween 20 (Product No. P1379) in 4x SSC.
- Blocking buffer: 5% bovine albumin (Product No. A3803) in detection buffer.
- Antibody or detection compound (e.g., Streptavidin-Cy3, Product No. S6402) in blocking buffer.
- DAPI (Product No. D9542) 2 µg/ml in antifade mounting medium.
- Fluorescence microscope, filters and optional triple band pass filter (x58, Omega Optics).
- Glass slides (Product No. S8400).
- Plastic cover slips for incubation and hybridization steps (cut from autoclavable waste bags, e.g., Product No. B4408).
- Heat block/ modified thermocycler.
- Coplin jars for washing steps (Product No. S6016, S5641 or S5891).
Procedure
Slide Preparation Hybridization Detection
Slide Preparation
- Start with chromosome preparations from any cell type.
- Incubate with 200 µl RNase for 1 hour at 37 °C
- Wash slides in 2x SSC for 5 minutes, repeat.
- Rinse slides in 10 mM HCl.
- Incubate with 200 µl pepsin for 10 minutes at 37 °C.
- Rinse slides in deionized H2O.
- Wash slides in 2x SSC for 5 minutes, repeat.
- Stabilize slides in paraformaldehyde for 10 minutes.
- Wash slides in 2x SSC for 5 minutes, repeat.
- Dehydrate slides in an ethanol series: 70%, 80%, 95%; 2 minutes each.
- Air dry.
Hybridization
- Prepare 30 µl hybridization solution per slide. Heat to 70 °C. for 10 minutes and place on ice.
- Place 30 µl of hybridization solution on each slide and cover with a plastic cover slip.
- Denature slide at 65-70 °C for 5 minutes on heat block.
- Gradually decrease temperature to 37 °C.
- Hybridize at 37 °C overnight in humidity chamber.
Detection
- Wash slides in 2x SSC to remove coverslip.
- Wash slides in wash buffer at 40 °C for 5 minutes, repeat.
- Wash slides in 0.1x SSC at 40 °C for 5-15 minutes.
- Wash slides in 2x SSC at 40 °C for 5-15 minutes.
- Cool slides to room temperature.
- Equilibrate slides in detection buffer for 5 minutes.
- Block in blocking buffer for 20-30 minutes.
- Incubate with 50 µl antibody or detection compound for 30-60 minutes (e.g., 5 µg/ml Streptavidin-Cy3 in blocking buffer).
- Wash slides in 2x SSC for 5 minutes, repeat twice.
- Counterstain with DAPI solution for 10 minutes.
- Rinse briefly and mount in antifade mounting medium.
- Analyze with fluorescence microscope.
References
- Heslop-Harrison, J.S., et al., Technique, 3, 109 (1991).
- Leitch, A.R., et al., in: In situ Hybridization, Bios Scientific Publishers, Oxford, England (1994).
- Schwarzacher, T., et al., in: Plant Cell Biology: A Practical Approach, Harris, N., and Oparka, K.J. (eds.), Oxford University Press, Oxford, England, pp. 127 (1994).
back to Antibodies Protocols back to top
|